Search
Search

| Assay Name: | hsa-miR-499a-5p |
|---|---|
| miRBase Accession Number: | MI0003183 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUU |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Bovine, Callithrix jacchus, Dog, Goat, Horse, Ictalurus punctatus, Monodelphis domestica, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-499a-5p | MIMAT0002870 |
| Mouse | mmu-miR-499-5p | MIMAT0003482 |
| Rat | rno-miR-499-5p | MIMAT0003381 |
| Alligator mississippiensis | ami-miR-499-5p | MIMAT0038301 |
| Bovine | bta-miR-499 | MIMAT0003536 |
| Callithrix jacchus | cja-miR-499a | MIMAT0039510 |
| View More Rows | ||
| Dog | cfa-miR-499 | MIMAT0006655 |
| Goat | chi-miR-499-5p | MIMAT0036261 |
| Horse | eca-miR-499-5p | MIMAT0013108 |
| Ictalurus punctatus | ipu-miR-499 | MIMAT0029521 |
| Monodelphis domestica | mdo-miR-499-5p | MIMAT0012753 |
| Pig | ssc-miR-499-5p | MIMAT0013877 |
| Pongo pygmaeus | ppy-miR-499-5p | MIMAT0015920 |
| Pteropus alecto | pal-miR-499a-5p | MIMAT0039820 |
| Rhesus monkey | mml-miR-499-5p | MIMAT0006361 |
| Tupaia chinensis | tch-miR-499a-5p | MIMAT0036509 |