Search
Search

| Assay Name: | hsa-miR-639 |
|---|---|
| miRBase Accession Number: | MI0003654 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUCGCUGCGGUUGCGAGCGCUGU |
| Species: | Human, Pongo pygmaeus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000549, mir-639 |
Mature miRNA Sequence: | AUCGCUGCGGUUGCGAGCGCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-639 | MIMAT0003309 |
| Pongo pygmaeus | ppy-miR-639 | MIMAT0016077 |