Search
Search

| Assay Name: | hsa-miR-614 |
|---|---|
| miRBase Accession Number: | MI0003627 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAACGCCUGUUCUUGCCAGGUGG |
| Species: | Human, Pan troglodytes, Pongo pygmaeus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000647, mir-614 |
Mature miRNA Sequence: | GAACGCCUGUUCUUGCCAGGUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-614 | MIMAT0003282 |
| Pan troglodytes | ptr-miR-614 | MIMAT0008276 |
| Pongo pygmaeus | ppy-miR-614 | MIMAT0016055 |