Search
Search

| Assay Name: | hsa-miR-23b* |
|---|---|
| miRBase Accession Number: | MI0000439 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGGUUCCUGGCAUGCUGAUUU |
| Species: | Human, Bovine, Goat, Ophiophagus hannah, Ornithorhynchus anatinus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | UGGGUUCCUGGCAUGCUGAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-23b-5p | MIMAT0004587 |
| Bovine | bta-miR-10174-5p | MIMAT0040927 |
| Goat | chi-miR-23b-5p | MIMAT0036088 |
| Ophiophagus hannah | oha-miR-23b-5p | MIMAT0036856 |
| Ornithorhynchus anatinus | oan-miR-23b-5p | MIMAT0006958 |