Search
Search

| Assay Name: | hsa-miR-339-5p |
|---|---|
| miRBase Accession Number: | MI0000815 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCUGUCCUCCAGGAGCUCACG |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000193, mir-339 |
Mature miRNA Sequence: | UCCCUGUCCUCCAGGAGCUCACG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-339-5p | MIMAT0000764 | |
| Mouse | mmu-miR-339-5p | MIMAT0000584 | |
| Rat | rno-miR-339-5p | MIMAT0000583 | |
| Cricetulus griseus | cgr-miR-339 | MIMAT0023919 | |
| Dasypus novemcinctus | dno-miR-339-5p | MIMAT0047821 | |
| Guinea pig | cpo-miR-339-5p | MIMAT0047176 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-339-5p | MIMAT0015842 | |
| Rhesus monkey | mml-miR-339-5p | MIMAT0006279 | |