Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481070_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-339-5p
          Stem-loop Accession Number: MI0000815
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 1022933 - 1023026 [-] on Build GRCh38
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Species: Human, Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481070_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000193, mir-339

          Mature miRNA Sequence:

          UCCCUGUCCUCCAGGAGCUCACG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-339-5p MIMAT0000764
          Mouse mmu-miR-339-5p MIMAT0000584
          Rat rno-miR-339-5p MIMAT0000583
          Cricetulus griseus cgr-miR-339 MIMAT0023919
          Dasypus novemcinctus dno-miR-339-5p MIMAT0047821
          Guinea pig cpo-miR-339-5p MIMAT0047176
          View More Rows
          Pongo pygmaeus ppy-miR-339-5p MIMAT0015842
          Rhesus monkey mml-miR-339-5p MIMAT0006279

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-339
          Stem-loop Accession # MI0000815
          Stem-loop Sequence
          CGGGGCGGCCGCUCUCCCUGUCCUCCAGGAGCUCACGUGUGCCUGCCUGUGAGCGCCUCGACGACAGAGCCGGCGCCUGCCCCAGUGUCUGCGC
          Chromosome Location Chr. 7 - 1022933 - 1023026 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000764
          miRBase ID: hsa-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: Chr. 7 - 1022933 - 1023026 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-339
          Stem-loop Accession # MI0000621
          Stem-loop Sequence
          ACGGGGUGGCCACUAUCCCUGUCCUCCAGGAGCUCACGUAUGCCUGCCUGUGAGCGCCUCGGCGACAGAGCCGGUGUCCACCCCUGCACUGUCCAC
          Chromosome Location Chr. 5 - 139369650 - 139369745 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000584
          miRBase ID: mmu-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: Chr. 5 - 139369650 - 139369745 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-339
          Stem-loop Accession # MI0000620
          Stem-loop Sequence
          ACGGGGUGGACACCGUCCCUGUCCUCCAGGAGCUCACGUAUGCCUGCCUGUGAGCGCCUCGACGACAGAGCCAGAGUCCACCCCUGCACUGCCCAA
          Chromosome Location Chr. 12 - 17369002 - 17369097 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000583
          miRBase ID: rno-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: Chr. 12 - 17369002 - 17369097 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-339
          Stem-loop Accession # MI0020500
          Stem-loop Sequence
          CAGCCACCGUCCCUGUCCUCCAGGAGCUCACGUAUGCCUGCCUGUGAGCACCUCGAUGACAGAGCCGGUGUCCACCCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023919
          miRBase ID: cgr-miR-339
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-339
          Stem-loop Accession # MI0039075
          Stem-loop Sequence
          UCCCUGUCCUCCAGGAGCUCACGUGGGUCAGGCUGUGAGCGCCUCGAGGGCAGAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047821
          miRBase ID: dno-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-339
          Stem-loop Accession # MI0038736
          Stem-loop Sequence
          UCCCUGUCCUCCAGGAGCUCACGUAUGCCUGCUUGUGAGCGCCUCGAUGACAGAGCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047176
          miRBase ID: cpo-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-339
          Stem-loop Accession # MI0014903
          Stem-loop Sequence
          CGGGGCGGCCGCUCUCCCUGUCCUCCAGGAGCUCACGUGUGCCUGCCUGUGAGCGCCUUCGACGACAGGAGCCGGCGCCCGCCCCAGUGUCUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015842
          miRBase ID: ppy-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-339
          Stem-loop Accession # MI0007702
          Stem-loop Sequence
          CGGGGCGGCCGCUCUCCCUGUCCUCCAGGAGCUCACGUGUGCCUGCCUGUGAGCGCCUCGACGACAGAGCCGGCGCCCGCCCCAGUGUCUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006279
          miRBase ID: mml-miR-339-5p
          Mature miRNA Sequence: UCCCUGUCCUCCAGGAGCUCACG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303641_pri
          TaqMan™ Pri-miRNA Assay : Rn03464972_pri
          TaqMan™ Pri-miRNA Assay : Mm03306988_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12347
          Ambion® Pre-miR™ miRNA Precursor : PM12347
          TaqMan™ MicroRNA Assay : 002257
          mirVana® miRNA inhibitor : MH12347
          mirVana® miRNA mimic : MC12347
          TaqMan™ Advanced miRNA Assay : rno481070_mir
          TaqMan™ Advanced miRNA Assay : 478040_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline