Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002296
          Assay Name: hsa-miR-885-5p
          miRBase Accession Number: MI0005560
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 10394489 - 10394562 [-] on Build GRCh38
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Species: Human, Bovine, Callithrix jacchus, Dasypus novemcinctus, Dog, Horse, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002296
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000532, mir-885

          Mature miRNA Sequence:

          UCCAUUACACUACCCUGCCUCU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-885-5p MIMAT0004947
          Bovine bta-miR-885 MIMAT0009382
          Callithrix jacchus cja-miR-885 MIMAT0039464
          Dasypus novemcinctus dno-miR-885-5p MIMAT0048020
          Dog cfa-miR-885 MIMAT0009933
          Horse eca-miR-885-5p MIMAT0013081
          View More Rows
          Pig ssc-miR-885-5p MIMAT0013902
          Pongo pygmaeus ppy-miR-885-5p MIMAT0016118
          Pteropus alecto pal-miR-885-5p MIMAT0040017
          Rabbit ocu-miR-885-5p MIMAT0048579
          Rhesus monkey mml-miR-885-5p MIMAT0006522

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-885
          Stem-loop Accession # MI0005560
          Stem-loop Sequence
          CCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location Chr. 3 - 10394489 - 10394562 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004947
          miRBase ID: hsa-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: Chr. 3 - 10394489 - 10394562 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-885
          Stem-loop Accession # MI0009904
          Stem-loop Sequence
          CCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCACGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009382
          miRBase ID: bta-miR-885
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-885
          Stem-loop Accession # MI0031988
          Stem-loop Sequence
          GGCCCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGUGGGGUGUAGUGGAUAGAGCACGGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039464
          miRBase ID: cja-miR-885
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-885
          Stem-loop Accession # MI0039177
          Stem-loop Sequence
          UCCAUUACACUACCCUGCCUCUUCUCCACGAGAGGCAGCUGGGUGUAGCGGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048020
          miRBase ID: dno-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-885
          Stem-loop Accession # MI0010432
          Stem-loop Sequence
          CCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009933
          miRBase ID: cfa-miR-885
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-885
          Stem-loop Accession # MI0012828
          Stem-loop Sequence
          CCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013081
          miRBase ID: eca-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-885
          Stem-loop Accession # MI0013119
          Stem-loop Sequence
          CACUCGGCCCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCACGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013902
          miRBase ID: ssc-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-885
          Stem-loop Accession # MI0015155
          Stem-loop Sequence
          CCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016118
          miRBase ID: ppy-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-885
          Stem-loop Accession # MI0032450
          Stem-loop Sequence
          UCCAUUACACUACCCUGCCUCUUCUCUGUGAGAGGCAGUGGGGUGUAGUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040017
          miRBase ID: pal-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-885
          Stem-loop Accession # MI0039472
          Stem-loop Sequence
          UCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048579
          miRBase ID: ocu-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-885
          Stem-loop Accession # MI0007919
          Stem-loop Sequence
          CCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006522
          miRBase ID: mml-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305150_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12960
          Ambion® Pre-miR™ miRNA Precursor : PM12960
          mirVana® miRNA inhibitor : MH12960
          mirVana® miRNA mimic : MC12960
          TaqMan™ Advanced miRNA Assay : 478207_mir
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline