Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12960
          Assay Name: hsa-miR-885-5p
          miRBase Accession Number: MI0005560
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 10394489 - 10394562 [-] on Build GRCh38
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Species: Human, Bovine, Callithrix jacchus, Dasypus novemcinctus, Dog, Horse, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12960

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000532, mir-885

          Mature miRNA Sequence:

          UCCAUUACACUACCCUGCCUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-885-5p MIMAT0004947
          Bovine bta-miR-885 MIMAT0009382
          Callithrix jacchus cja-miR-885 MIMAT0039464
          Dasypus novemcinctus dno-miR-885-5p MIMAT0048020
          Dog cfa-miR-885 MIMAT0009933
          Horse eca-miR-885-5p MIMAT0013081
          View More Rows
          Pig ssc-miR-885-5p MIMAT0013902
          Pongo pygmaeus ppy-miR-885-5p MIMAT0016118
          Pteropus alecto pal-miR-885-5p MIMAT0040017
          Rabbit ocu-miR-885-5p MIMAT0048579
          Rhesus monkey mml-miR-885-5p MIMAT0006522

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-885
          Stem-loop Accession # MI0005560
          Stem-loop Sequence
          CCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location Chr. 3 - 10394489 - 10394562 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004947
          miRBase ID: hsa-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: Chr. 3 - 10394489 - 10394562 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-885
          Stem-loop Accession # MI0009904
          Stem-loop Sequence
          CCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCACGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009382
          miRBase ID: bta-miR-885
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-885
          Stem-loop Accession # MI0031988
          Stem-loop Sequence
          GGCCCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGUGGGGUGUAGUGGAUAGAGCACGGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039464
          miRBase ID: cja-miR-885
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-885
          Stem-loop Accession # MI0039177
          Stem-loop Sequence
          UCCAUUACACUACCCUGCCUCUUCUCCACGAGAGGCAGCUGGGUGUAGCGGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048020
          miRBase ID: dno-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-885
          Stem-loop Accession # MI0010432
          Stem-loop Sequence
          CCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009933
          miRBase ID: cfa-miR-885
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-885
          Stem-loop Accession # MI0012828
          Stem-loop Sequence
          CCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013081
          miRBase ID: eca-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-885
          Stem-loop Accession # MI0013119
          Stem-loop Sequence
          CACUCGGCCCGCGCUCUCUCCAUUACACUACCCUGCCUCUUCUCCACGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013902
          miRBase ID: ssc-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-885
          Stem-loop Accession # MI0015155
          Stem-loop Sequence
          CCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016118
          miRBase ID: ppy-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-885
          Stem-loop Accession # MI0032450
          Stem-loop Sequence
          UCCAUUACACUACCCUGCCUCUUCUCUGUGAGAGGCAGUGGGGUGUAGUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040017
          miRBase ID: pal-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-885
          Stem-loop Accession # MI0039472
          Stem-loop Sequence
          UCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048579
          miRBase ID: ocu-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-885
          Stem-loop Accession # MI0007919
          Stem-loop Sequence
          CCGCACUCUCUCCAUUACACUACCCUGCCUCUUCUCCAUGAGAGGCAGCGGGGUGUAGUGGAUAGAGCACGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006522
          miRBase ID: mml-miR-885-5p
          Mature miRNA Sequence: UCCAUUACACUACCCUGCCUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305150_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12960
          Ambion® Pre-miR™ miRNA Precursor : PM12960
          TaqMan™ MicroRNA Assay : 002296
          mirVana® miRNA mimic : MC12960
          TaqMan™ Advanced miRNA Assay : 478207_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-ncdd4:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline