Search
Search

| Assay Name: | hsa-miR-1237 |
|---|---|
| miRBase Accession Number: | MI0006327 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCUUCUGCUCCGUCCCCCAG |
| Species: | Human, Pan troglodytes |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000585, mir-1237 |
Mature miRNA Sequence: | UCCUUCUGCUCCGUCCCCCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1237-3p | MIMAT0005592 |
| Pan troglodytes | ptr-miR-1237 | MIMAT0007969 |