Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 008345_mat
          Assay Name: aae-miR-981
          miRBase Accession Number: MI0005838
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Species: Drosophila melanogaster, Aedes aegypti, Bactrocera dorsalis, Culex quinquefasciatus, Daphnia pulex, Dinoponera quadriceps, Drosophila pseudoobscura, Drosophila virilis, Heliconius melpomene, Manduca sexta, Polistes canadensis, Tribolium castaneum
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 008345_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000710, mir-981

          Mature miRNA Sequence:

          UUCGUUGUCGACGAAACCUGCA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Drosophila melanogaster dme-miR-981-3p MIMAT0005497
          Aedes aegypti aae-miR-981 MIMAT0014267
          Bactrocera dorsalis bdo-miR-981 MIMAT0045399
          Culex quinquefasciatus cqu-miR-981 MIMAT0014386
          Daphnia pulex dpu-miR-981 MIMAT0012675
          Dinoponera quadriceps dqu-miR-981-3p MIMAT0045760
          View More Rows
          Drosophila pseudoobscura dps-miR-981-3p MIMAT0012220
          Drosophila virilis dvi-miR-981-3p MIMAT0035598
          Heliconius melpomene hme-miR-981 MIMAT0024999
          Manduca sexta mse-miR-981 MIMAT0024433
          Polistes canadensis pca-miR-981-3p MIMAT0045926
          Tribolium castaneum tca-miR-981 MIMAT0010098

          Stem-loop Details



          Drosophila melanogaster

          Stem-loop ID dme-mir-981
          Stem-loop Accession # MI0005838
          Stem-loop Sequence
          AAACAUCCUCACUGAAGUCGGGUUUCGUUAGCAGCGGGCUGUUUUAAUAAAUUCAACAAGUUCGUUGUCGACGAAACCUGCAUGCUGUGUGGAAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005497
          miRBase ID: dme-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Aedes aegypti

          Stem-loop ID aae-mir-981
          Stem-loop Accession # MI0013473
          Stem-loop Sequence
          AAACCUUGCCUGGAUCGGGUUUCGCGGCGAACGGGCUUCCACCAUGAUGAAUAGCAGUUCGUUGUCGACGAAACCUGCACAUUGCAAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014267
          miRBase ID: aae-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Bactrocera dorsalis

          Stem-loop ID bdo-mir-981
          Stem-loop Accession # MI0037276
          Stem-loop Sequence
          CGGGUUUCGUAAGCAGCGGGCUGUUAAACUGUUAAAACGAGUUCGUUGUCGACGAAACCUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045399
          miRBase ID: bdo-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Culex quinquefasciatus

          Stem-loop ID cqu-mir-981
          Stem-loop Accession # MI0013584
          Stem-loop Sequence
          AAACACCUUGUCUGGAUCGGGUUUCGCGGCAAACGGGCUGUACUGAUUCGUGAAAAAUCAGUUCGUUGUCGACGAAACCUGCACACUGCAAGGUUGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014386
          miRBase ID: cqu-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Daphnia pulex

          Stem-loop ID dpu-mir-981
          Stem-loop Accession # MI0012256
          Stem-loop Sequence
          AAUUGUGCCGUGCUUGUCCGGGUUUCGCGACGCUCGAACGCCGUUUACUUUCACAGUCCGUUCGUUGUCGACGAAACCUGCAUAUCGCACGGUCACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012675
          miRBase ID: dpu-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Dinoponera quadriceps

          Stem-loop ID dqu-mir-981
          Stem-loop Accession # MI0037530
          Stem-loop Sequence
          CAGAAGGAGGAGCCAAGUAUGGACGCAGACGUCCGGGUUUCGUGACACGCGAACCGUUUGUAAAACUUGGUUCGUUGUCGACGAAACCUGCACGAUCUGCAACCACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045760
          miRBase ID: dqu-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Drosophila pseudoobscura

          Stem-loop ID dps-mir-981
          Stem-loop Accession # MI0011592
          Stem-loop Sequence
          UCCACAUUGAAAUCGGGUUUCGUUAACAGCGGGCUGUUAUGAAUAAAUCAACAAGUUCGUUGUCGACGAAACCUGCAUGCUGUGUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012220
          miRBase ID: dps-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Drosophila virilis

          Stem-loop ID dvi-mir-981
          Stem-loop Accession # MI0029266
          Stem-loop Sequence
          CUUUAUUUAACAAUCUACAUUGAACUCAGAUUUCGUCAGCAGCGGGCUGUGAUGAUUAUGAACAAGUUCGUUGUCGACGAAACCUGCAUACUGUGUGGAGUGGCAACAAGUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035598
          miRBase ID: dvi-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Heliconius melpomene

          Stem-loop ID hme-mir-981
          Stem-loop Accession # MI0021796
          Stem-loop Sequence
          AGAAACCGCACAUAAUUCGGGUUUCGAGACGAUGAAACCGUAUGUACAAUUGGUUCGUUGUCGACGAAACCUGCAAAAUGUGCGGCGAACUCCAACUGGAAAAAAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024999
          miRBase ID: hme-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Manduca sexta

          Stem-loop ID mse-mir-981
          Stem-loop Accession # MI0021023
          Stem-loop Sequence
          GAAAAUGAAGAAGAAUCCGCACAUGAUUCGGGUUUCGAGACGAUGAAACCGUGAUUAAAUUAAAUGGUUCGUUGUCGACGAAACCUGCAAAAUGUGCGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024433
          miRBase ID: mse-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Polistes canadensis

          Stem-loop ID pca-mir-981
          Stem-loop Accession # MI0037616
          Stem-loop Sequence
          AGUAAAAUAUACAAUUAUUGGACGCAGACGUCCGGGUUUCGUGACAGGCGAGCCGUUUCAUUAAACUUGGUUCGUUGUCGACGAAACCUGCACGAUCUAUGACCGCAUCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045926
          miRBase ID: pca-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Tribolium castaneum

          Stem-loop ID tca-mir-981
          Stem-loop Accession # MI0010592
          Stem-loop Sequence
          UACAAGACUUAUAUCUCAUAAGUUACGGGUUUCGGGGCAUCGGAACCGAUUUUAAGCUGGUUCGUUGUCGACGAAACCUGCAGACUAUGGGACACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010098
          miRBase ID: tca-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH13765
          mirVana® miRNA mimic : MC13765


          miRBase Alias:

          Derxomyces pseudoschimicola: dps-miR-981 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline