Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13765
          Assay Name: dme-miR-981-3p
          miRBase Accession Number: MI0005838
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Species: Drosophila melanogaster, Aedes aegypti, Bactrocera dorsalis, Culex quinquefasciatus, Daphnia pulex, Dinoponera quadriceps, Drosophila pseudoobscura, Drosophila virilis, Heliconius melpomene, Manduca sexta, Polistes canadensis, Tribolium castaneum
          Product Type: mirVana® miRNA mimic
          Assay ID MC13765

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000710, mir-981

          Mature miRNA Sequence:

          UUCGUUGUCGACGAAACCUGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Drosophila melanogaster dme-miR-981-3p MIMAT0005497
          Aedes aegypti aae-miR-981 MIMAT0014267
          Bactrocera dorsalis bdo-miR-981 MIMAT0045399
          Culex quinquefasciatus cqu-miR-981 MIMAT0014386
          Daphnia pulex dpu-miR-981 MIMAT0012675
          Dinoponera quadriceps dqu-miR-981-3p MIMAT0045760
          View More Rows
          Drosophila pseudoobscura dps-miR-981-3p MIMAT0012220
          Drosophila virilis dvi-miR-981-3p MIMAT0035598
          Heliconius melpomene hme-miR-981 MIMAT0024999
          Manduca sexta mse-miR-981 MIMAT0024433
          Polistes canadensis pca-miR-981-3p MIMAT0045926
          Tribolium castaneum tca-miR-981 MIMAT0010098

          Stem-loop Details



          Drosophila melanogaster

          Stem-loop ID dme-mir-981
          Stem-loop Accession # MI0005838
          Stem-loop Sequence
          AAACAUCCUCACUGAAGUCGGGUUUCGUUAGCAGCGGGCUGUUUUAAUAAAUUCAACAAGUUCGUUGUCGACGAAACCUGCAUGCUGUGUGGAAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005497
          miRBase ID: dme-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Aedes aegypti

          Stem-loop ID aae-mir-981
          Stem-loop Accession # MI0013473
          Stem-loop Sequence
          AAACCUUGCCUGGAUCGGGUUUCGCGGCGAACGGGCUUCCACCAUGAUGAAUAGCAGUUCGUUGUCGACGAAACCUGCACAUUGCAAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014267
          miRBase ID: aae-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Bactrocera dorsalis

          Stem-loop ID bdo-mir-981
          Stem-loop Accession # MI0037276
          Stem-loop Sequence
          CGGGUUUCGUAAGCAGCGGGCUGUUAAACUGUUAAAACGAGUUCGUUGUCGACGAAACCUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045399
          miRBase ID: bdo-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Culex quinquefasciatus

          Stem-loop ID cqu-mir-981
          Stem-loop Accession # MI0013584
          Stem-loop Sequence
          AAACACCUUGUCUGGAUCGGGUUUCGCGGCAAACGGGCUGUACUGAUUCGUGAAAAAUCAGUUCGUUGUCGACGAAACCUGCACACUGCAAGGUUGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014386
          miRBase ID: cqu-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Daphnia pulex

          Stem-loop ID dpu-mir-981
          Stem-loop Accession # MI0012256
          Stem-loop Sequence
          AAUUGUGCCGUGCUUGUCCGGGUUUCGCGACGCUCGAACGCCGUUUACUUUCACAGUCCGUUCGUUGUCGACGAAACCUGCAUAUCGCACGGUCACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012675
          miRBase ID: dpu-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Dinoponera quadriceps

          Stem-loop ID dqu-mir-981
          Stem-loop Accession # MI0037530
          Stem-loop Sequence
          CAGAAGGAGGAGCCAAGUAUGGACGCAGACGUCCGGGUUUCGUGACACGCGAACCGUUUGUAAAACUUGGUUCGUUGUCGACGAAACCUGCACGAUCUGCAACCACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045760
          miRBase ID: dqu-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Drosophila pseudoobscura

          Stem-loop ID dps-mir-981
          Stem-loop Accession # MI0011592
          Stem-loop Sequence
          UCCACAUUGAAAUCGGGUUUCGUUAACAGCGGGCUGUUAUGAAUAAAUCAACAAGUUCGUUGUCGACGAAACCUGCAUGCUGUGUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012220
          miRBase ID: dps-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Drosophila virilis

          Stem-loop ID dvi-mir-981
          Stem-loop Accession # MI0029266
          Stem-loop Sequence
          CUUUAUUUAACAAUCUACAUUGAACUCAGAUUUCGUCAGCAGCGGGCUGUGAUGAUUAUGAACAAGUUCGUUGUCGACGAAACCUGCAUACUGUGUGGAGUGGCAACAAGUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035598
          miRBase ID: dvi-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Heliconius melpomene

          Stem-loop ID hme-mir-981
          Stem-loop Accession # MI0021796
          Stem-loop Sequence
          AGAAACCGCACAUAAUUCGGGUUUCGAGACGAUGAAACCGUAUGUACAAUUGGUUCGUUGUCGACGAAACCUGCAAAAUGUGCGGCGAACUCCAACUGGAAAAAAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024999
          miRBase ID: hme-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Manduca sexta

          Stem-loop ID mse-mir-981
          Stem-loop Accession # MI0021023
          Stem-loop Sequence
          GAAAAUGAAGAAGAAUCCGCACAUGAUUCGGGUUUCGAGACGAUGAAACCGUGAUUAAAUUAAAUGGUUCGUUGUCGACGAAACCUGCAAAAUGUGCGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024433
          miRBase ID: mse-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Polistes canadensis

          Stem-loop ID pca-mir-981
          Stem-loop Accession # MI0037616
          Stem-loop Sequence
          AGUAAAAUAUACAAUUAUUGGACGCAGACGUCCGGGUUUCGUGACAGGCGAGCCGUUUCAUUAAACUUGGUUCGUUGUCGACGAAACCUGCACGAUCUAUGACCGCAUCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045926
          miRBase ID: pca-miR-981-3p
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA

          Tribolium castaneum

          Stem-loop ID tca-mir-981
          Stem-loop Accession # MI0010592
          Stem-loop Sequence
          UACAAGACUUAUAUCUCAUAAGUUACGGGUUUCGGGGCAUCGGAACCGAUUUUAAGCUGGUUCGUUGUCGACGAAACCUGCAGACUAUGGGACACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010098
          miRBase ID: tca-miR-981
          Mature miRNA Sequence: UUCGUUGUCGACGAAACCUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 008345_mat
          mirVana® miRNA inhibitor : MH13765


          miRBase Alias:

          Drosophila melanogaster: dme-miR-981 (v16)
          Derxomyces pseudoschimicola: dps-miR-981 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline