Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 243584_mat
          Assay Name: hsa-miR-3074
          miRBase Accession Number: MI0014181
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 95086014 - 95086094 [-] on Build GRCh38
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Species: Human, Mouse, Rat, Artibeus jamaicensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 243584_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0001103, mir-3074

          Mature miRNA Sequence:

          GAUAUCAGCUCAGUAGGCACCG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3074-3p MIMAT0015027
          Mouse mmu-miR-3074-1-3p MIMAT0014857
          Rat rno-miR-3074 MIMAT0017815
          Artibeus jamaicensis aja-miR-3074 MIMAT0031145

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3074
          Stem-loop Accession # MI0014181
          Stem-loop Sequence
          GCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUAAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGU
          Chromosome Location Chr. 9 - 95086014 - 95086094 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0015027
          miRBase ID: hsa-miR-3074-3p
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 9 - 95086014 - 95086094 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-3074-1
          Stem-loop Accession # MI0014037
          Stem-loop Sequence
          GGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUGAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCC
          Chromosome Location Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014857
          miRBase ID: mmu-miR-3074-1-3p
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-3074
          Stem-loop Accession # MI0015411
          Stem-loop Sequence
          GCUUUUUGCUCAAGGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUGAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCCAAUCACAGCUGGGCAGUGC
          Chromosome Location Chr. 17 - 823940 - 824056 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017815
          miRBase ID: rno-miR-3074
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 17 - 823940 - 824056 [-] on Build Rnor_6.0

          Artibeus jamaicensis

          Stem-loop ID aja-mir-3074
          Stem-loop Accession # MI0024367
          Stem-loop Sequence
          GGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUAAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031145
          miRBase ID: aja-miR-3074
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465740_pri
          TaqMan™ Pri-miRNA Assay : Hs03303056_pri
          TaqMan™ Pri-miRNA Assay : Mm03306468_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM18636
          Ambion® Pre-miR™ miRNA Precursor : PM18636
          mirVana® miRNA inhibitor : MH18636
          mirVana® miRNA mimic : MC18636
          TaqMan™ Advanced miRNA Assay : 478803_mir
          TaqMan™ Advanced miRNA Assay : mmu481714_mir
          TaqMan™ Advanced miRNA Assay : rno478803_mir


          miRBase Alias:

          Human: hsa-miR-3074 (v16)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline