Search
Search

| Assay Name: | ssc-miR-335 |
|---|---|
| miRBase Accession Number: | MI0031980 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAAGAGCAAUAACGAAAAAUG |
| Species: | Callithrix jacchus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Pig, Pteropus alecto, Rabbit, Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UCAAGAGCAAUAACGAAAAAUG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Callithrix jacchus | cja-miR-335-5p | MIMAT0039456 | |
| Dasypus novemcinctus | dno-miR-335-5p | MIMAT0047815 | |
| Gorilla gorilla | ggo-miR-335 | MIMAT0024122 | |
| Guinea pig | cpo-miR-335-5p | MIMAT0047170 | |
| Pig | ssc-miR-335 | MIMAT0013955 | |
| Pteropus alecto | pal-miR-335-5p | MIMAT0040161 | |
| View More Rows | |||
| Rabbit | ocu-miR-335-5p | MIMAT0048382 | |
| Tupaia chinensis | tch-miR-335-5p | MIMAT0036618 | |