Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH16607
          Assay Name: cja-miR-335-5p
          miRBase Accession Number: MI0031980
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Species: Callithrix jacchus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Pig, Pteropus alecto, Rabbit, Tupaia chinensis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH16607

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          UCAAGAGCAAUAACGAAAAAUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Callithrix jacchus cja-miR-335-5p MIMAT0039456
          Dasypus novemcinctus dno-miR-335-5p MIMAT0047815
          Gorilla gorilla ggo-miR-335 MIMAT0024122
          Guinea pig cpo-miR-335-5p MIMAT0047170
          Pig ssc-miR-335 MIMAT0013955
          Pteropus alecto pal-miR-335-5p MIMAT0040161
          View More Rows
          Rabbit ocu-miR-335-5p MIMAT0048382
          Tupaia chinensis tch-miR-335-5p MIMAT0036618

          Stem-loop Details



          Callithrix jacchus

          Stem-loop ID cja-mir-335
          Stem-loop Accession # MI0031980
          Stem-loop Sequence
          UGAGCGGGGGUCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACCUCCUCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039456
          miRBase ID: cja-miR-335-5p
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-335
          Stem-loop Accession # MI0039072
          Stem-loop Sequence
          UCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047815
          miRBase ID: dno-miR-335-5p
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-335
          Stem-loop Accession # MI0020669
          Stem-loop Sequence
          GGAAAUGAUUUGAGCGGGGGUCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACCUCCUCUCAUUUGCUAUAUUCAAUUAAGUAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024122
          miRBase ID: ggo-miR-335
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-335
          Stem-loop Accession # MI0038733
          Stem-loop Sequence
          UCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047170
          miRBase ID: cpo-miR-335-5p
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-335
          Stem-loop Accession # MI0013165
          Stem-loop Sequence
          CUUUUGGGCGGGGGUCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACCUCCUCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013955
          miRBase ID: ssc-miR-335
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-335
          Stem-loop Accession # MI0032526
          Stem-loop Sequence
          UCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040161
          miRBase ID: pal-miR-335-5p
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-335
          Stem-loop Accession # MI0039371
          Stem-loop Sequence
          UCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048382
          miRBase ID: ocu-miR-335-5p
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-335
          Stem-loop Accession # MI0031291
          Stem-loop Sequence
          UCAAGAGCAAUAACGAAAAAUGUUUGUCAUAAACCGUUUUUCAUUAUUGCUCCUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036618
          miRBase ID: tch-miR-335-5p
          Mature miRNA Sequence: UCAAGAGCAAUAACGAAAAAUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 244560_mat
          mirVana® miRNA mimic : MC16607

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline