Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 245309_mat
          Assay Name: age-miR-29a
          miRBase Accession Number: MI0002658
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Species: Ateles geoffroyi, Gorilla gorilla, Lagothrix lagotricha, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Saguinus labiatus, Saimiri boliviensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 245309_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          CUAGCACCAUCUGAAAUCGGUU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Ateles geoffroyi age-miR-29a MIMAT0002361
          Gorilla gorilla ggo-miR-29a MIMAT0002360
          Lagothrix lagotricha lla-miR-29a MIMAT0002367
          Macaca nemestrina mne-miR-29a MIMAT0002368
          Pan paniscus ppa-miR-29a MIMAT0002362
          Pan troglodytes ptr-miR-29a MIMAT0002364
          View More Rows
          Pongo pygmaeus ppy-miR-29a MIMAT0002363
          Rhesus monkey mml-miR-29a-3p MIMAT0002365
          Saguinus labiatus sla-miR-29a MIMAT0002366
          Saimiri boliviensis sbo-miR-29a MIMAT0049505

          Stem-loop Details



          Ateles geoffroyi

          Stem-loop ID age-mir-29a
          Stem-loop Accession # MI0002658
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002361
          miRBase ID: age-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-29a
          Stem-loop Accession # MI0002657
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002360
          miRBase ID: ggo-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-29a
          Stem-loop Accession # MI0002664
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002367
          miRBase ID: lla-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-29a
          Stem-loop Accession # MI0002665
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002368
          miRBase ID: mne-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-29a
          Stem-loop Accession # MI0002659
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002362
          miRBase ID: ppa-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-29a
          Stem-loop Accession # MI0002661
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002364
          miRBase ID: ptr-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-29a
          Stem-loop Accession # MI0002660
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002363
          miRBase ID: ppy-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-29a
          Stem-loop Accession # MI0002662
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002365
          miRBase ID: mml-miR-29a-3p
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-29a
          Stem-loop Accession # MI0002663
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002366
          miRBase ID: sla-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-29a
          Stem-loop Accession # MI0040270
          Stem-loop Sequence
          CUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049505
          miRBase ID: sbo-miR-29a
          Mature miRNA Sequence: CUAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH11092
          mirVana® miRNA mimic : MC11092


          miRBase Alias:

          Macaca mulatta: mml-miR-29a (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline