Search
Search

| Assay Name: | age-miR-29a |
|---|---|
| miRBase Accession Number: | MI0002658 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAGCACCAUCUGAAAUCGGUU |
| Species: | Ateles geoffroyi, Gorilla gorilla, Lagothrix lagotricha, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Saguinus labiatus, Saimiri boliviensis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | CUAGCACCAUCUGAAAUCGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ateles geoffroyi | age-miR-29a | MIMAT0002361 |
| Gorilla gorilla | ggo-miR-29a | MIMAT0002360 |
| Lagothrix lagotricha | lla-miR-29a | MIMAT0002367 |
| Macaca nemestrina | mne-miR-29a | MIMAT0002368 |
| Pan paniscus | ppa-miR-29a | MIMAT0002362 |
| Pan troglodytes | ptr-miR-29a | MIMAT0002364 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-29a | MIMAT0002363 |
| Rhesus monkey | mml-miR-29a-3p | MIMAT0002365 |
| Saguinus labiatus | sla-miR-29a | MIMAT0002366 |
| Saimiri boliviensis | sbo-miR-29a | MIMAT0049505 |