Search
Search

| Assay Name: | mmu-miR-383* |
|---|---|
| miRBase Accession Number: | MI0000800 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCACAGCACUGCCUGGUCAGA |
| Species: | Mouse, Rat, Chicken, Dasypus novemcinctus, Ophiophagus hannah, Pig, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000137, mir-383 |
Mature miRNA Sequence: | CCACAGCACUGCCUGGUCAGA |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-383-3p | MIMAT0017082 | |
| Rat | rno-miR-383-3p | MIMAT0017197 | |
| Chicken | gga-miR-383-3p | MIMAT0026631 | |
| Dasypus novemcinctus | dno-miR-383-3p | MIMAT0047869 | |
| Ophiophagus hannah | oha-miR-383-3p | MIMAT0036920 | |
| Pig | ssc-miR-383 | MIMAT0017972 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-383-3p | MIMAT0040172 | |
| Python bivittatus | pbv-miR-383-3p | MIMAT0039047 | |
| Rabbit | ocu-miR-383-3p | MIMAT0048430 | |
| Taeniopygia guttata | tgu-miR-383-3p | MIMAT0026989 | |