Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481150_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-383-3p
          Stem-loop Accession Number: MI0000800
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 38252133 - 38252202 [-] on Build GRCm38
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Species: Mouse, Rat, Chicken, Dasypus novemcinctus, Ophiophagus hannah, Pig, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481150_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000137, mir-383

          Mature miRNA Sequence:

          CCACAGCACUGCCUGGUCAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-383-3p MIMAT0017082
          Rat rno-miR-383-3p MIMAT0017197
          Chicken gga-miR-383-3p MIMAT0026631
          Dasypus novemcinctus dno-miR-383-3p MIMAT0047869
          Ophiophagus hannah oha-miR-383-3p MIMAT0036920
          Pig ssc-miR-383 MIMAT0017972
          View More Rows
          Pteropus alecto pal-miR-383-3p MIMAT0040172
          Python bivittatus pbv-miR-383-3p MIMAT0039047
          Rabbit ocu-miR-383-3p MIMAT0048430
          Taeniopygia guttata tgu-miR-383-3p MIMAT0026989

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-383
          Stem-loop Accession # MI0000800
          Stem-loop Sequence
          CUCAGAUCAGAAGGUGACUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location Chr. 8 - 38252133 - 38252202 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017082
          miRBase ID: mmu-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: Chr. 8 - 38252133 - 38252202 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-383
          Stem-loop Accession # MI0003478
          Stem-loop Sequence
          UCCUCAGAUCAGAAGGUGACUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAGCA
          Chromosome Location Chr. 16 - 57519675 - 57519748 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017197
          miRBase ID: rno-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: Chr. 16 - 57519675 - 57519748 [+] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-mir-383
          Stem-loop Accession # MI0003706
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUAAAUAUUGAGCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026631
          miRBase ID: gga-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-383
          Stem-loop Accession # MI0039099
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUACUAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047869
          miRBase ID: dno-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-383
          Stem-loop Accession # MI0031467
          Stem-loop Sequence
          ACCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGUUUGAUAUGAAACAGCCACAGCACUGCCUGGUCAGAAAGAGCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036920
          miRBase ID: oha-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-383
          Stem-loop Accession # MI0015927
          Stem-loop Sequence
          CCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUAGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAGCAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017972
          miRBase ID: ssc-miR-383
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-383
          Stem-loop Accession # MI0032532
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040172
          miRBase ID: pal-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-383
          Stem-loop Accession # MI0030307
          Stem-loop Sequence
          CAGUCACCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGUUUGAUAUGAAACAGCCACAGCACUGCCUGGUCAGAAAGAGCAAGUGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039047
          miRBase ID: pbv-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-383
          Stem-loop Accession # MI0039396
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAGCAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048430
          miRBase ID: ocu-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-383
          Stem-loop Accession # MI0013744
          Stem-loop Sequence
          CCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGAGUAAAUAUUAAGCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026989
          miRBase ID: tgu-miR-383-3p
          Mature miRNA Sequence: CCACAGCACUGCCUGGUCAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307389_pri
          TaqMan™ Pri-miRNA Assay : Rn03465858_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11171
          Ambion® Pre-miR™ miRNA Precursor : PM11171
          TaqMan™ MicroRNA Assay : 463336_mat
          mirVana® miRNA inhibitor : MH11171
          mirVana® miRNA mimic : MC11171
          TaqMan™ Advanced miRNA Assay : rno481150_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline