Search
Search

| Assay Name: | mmu-miR-32* |
|---|---|
| miRBase Accession Number: | MI0000691 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAUUUAGUGUGUGUGAUAUU |
| Species: | Mouse, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000069, mir-32 |
Mature miRNA Sequence: | CAAUUUAGUGUGUGUGAUAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-32-3p | MIMAT0017050 |
| Cricetulus griseus | cgr-miR-32-3p | MIMAT0023905 |
| Dasypus novemcinctus | dno-miR-32-3p | MIMAT0047570 |
| Guinea pig | cpo-miR-32-3p | MIMAT0046917 |
| Rabbit | ocu-miR-32-3p | MIMAT0048164 |