Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH20030
          Assay Name: mmu-miR-32-3p
          miRBase Accession Number: MI0000691
          miRBase Version: v22.1
          Chromosome Location: Chr.4: 56895229 - 56895298 [-] on Build GRCm38
          Mature miRNA Sequence: CAAUUUAGUGUGUGUGAUAUU
          Species: Mouse, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Rabbit
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH20030

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000069, mir-32

          Mature miRNA Sequence:

          CAAUUUAGUGUGUGUGAUAUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-32-3p MIMAT0017050
          Cricetulus griseus cgr-miR-32-3p MIMAT0023905
          Dasypus novemcinctus dno-miR-32-3p MIMAT0047570
          Guinea pig cpo-miR-32-3p MIMAT0046917
          Rabbit ocu-miR-32-3p MIMAT0048164

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-32
          Stem-loop Accession # MI0000691
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017050
          miRBase ID: mmu-miR-32-3p
          Mature miRNA Sequence: CAAUUUAGUGUGUGUGAUAUU
          Chromosome Location: Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-32
          Stem-loop Accession # MI0020490
          Stem-loop Sequence
          CUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUUUUCACGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023905
          miRBase ID: cgr-miR-32-3p
          Mature miRNA Sequence: CAAUUUAGUGUGUGUGAUAUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-32
          Stem-loop Accession # MI0038941
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047570
          miRBase ID: dno-miR-32-3p
          Mature miRNA Sequence: CAAUUUAGUGUGUGUGAUAUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-32
          Stem-loop Accession # MI0038597
          Stem-loop Sequence
          UAUUGCACAUUUCUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046917
          miRBase ID: cpo-miR-32-3p
          Mature miRNA Sequence: CAAUUUAGUGUGUGUGAUAUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-32
          Stem-loop Accession # MI0039254
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048164
          miRBase ID: ocu-miR-32-3p
          Mature miRNA Sequence: CAAUUUAGUGUGUGUGAUAUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307087_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20030
          Ambion® Pre-miR™ miRNA Precursor : PM20030
          TaqMan™ MicroRNA Assay : 463590_mat
          mirVana® miRNA mimic : MC20030
          TaqMan™ Advanced miRNA Assay : mmu482741_mir


          miRBase Alias:

          Mouse: mmu-miR-32* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline