Search
Search

| Assay Name: | oar-miR-329b-5p |
|---|---|
| miRBase Accession Number: | MI0001725 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAGGUUUUCUGGGUUUCUGUUUC |
| Species: | Human, Dasypus novemcinctus, Rhesus monkey, Sheep |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | GAGGUUUUCUGGGUUUCUGUUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-329-5p | MIMAT0026555 |
| Dasypus novemcinctus | dno-miR-329-5p | MIMAT0047809 |
| Rhesus monkey | mml-miR-329-1-5p | MIMAT0026852 |
| Sheep | oar-miR-329b-5p | MIMAT0019263 |