Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478829_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-329-5p
          Stem-loop Accession Number: MI0001725
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101026785 - 101026864 [+] on Build GRCh38
          Mature miRNA Sequence: GAGGUUUUCUGGGUUUCUGUUUC
          Species: Human, Dasypus novemcinctus, Rhesus monkey, Sheep
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478829_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000110, mir-329

          Mature miRNA Sequence:

          GAGGUUUUCUGGGUUUCUGUUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-329-5p MIMAT0026555
          Dasypus novemcinctus dno-miR-329-5p MIMAT0047809
          Rhesus monkey mml-miR-329-1-5p MIMAT0026852
          Sheep oar-miR-329b-5p MIMAT0019263

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-329-1
          Stem-loop Accession # MI0001725
          Stem-loop Sequence
          GGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGACGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location Chr. 14 - 101026785 - 101026864 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026555
          miRBase ID: hsa-miR-329-5p
          Mature miRNA Sequence: GAGGUUUUCUGGGUUUCUGUUUC
          Chromosome Location: Chr. 14 - 101027100 - 101027183 [+] on Build GRCh38

          Stem-loop ID hsa-mir-329-2
          Stem-loop Accession # MI0001726
          Stem-loop Sequence
          GUGGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAUUGAGGACGAAACACACCUGGUUAACCUCUUUUCCAGUAUCAA
          Chromosome Location Chr. 14 - 101027100 - 101027183 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026555
          miRBase ID: hsa-miR-329-5p
          Mature miRNA Sequence: GAGGUUUUCUGGGUUUCUGUUUC
          Chromosome Location: Chr. 14 - 101027100 - 101027183 [+] on Build GRCh38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-329
          Stem-loop Accession # MI0039069
          Stem-loop Sequence
          GAGGUUUUCUGGGUUUCUGUUUCUUUAUUGAUGAUGAAACACACCUGGUUAACCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047809
          miRBase ID: dno-miR-329-5p
          Mature miRNA Sequence: GAGGUUUUCUGGGUUUCUGUUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-329-1
          Stem-loop Accession # MI0007695
          Stem-loop Sequence
          GUGGUACCUGAAGGGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026852
          miRBase ID: mml-miR-329-1-5p
          Mature miRNA Sequence: GAGGUUUUCUGGGUUUCUGUUUC
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-329b
          Stem-loop Accession # MI0016925
          Stem-loop Sequence
          CACUGGUGUUCUGGUCAGUGUUCACCCAUGGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUCAAAUCCCAUCUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019263
          miRBase ID: oar-miR-329b-5p
          Mature miRNA Sequence: GAGGUUUUCUGGGUUUCUGUUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303752_pri
          TaqMan™ Pri-miRNA Assay : Hs04225947_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20755
          Ambion® Pre-miR™ miRNA Precursor : PM20755
          TaqMan™ MicroRNA Assay : 463673_mat
          mirVana® miRNA inhibitor : MH20755
          mirVana® miRNA mimic : MC20755

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline