Search
Search

| Assay Name: | rno-miR-3552 |
|---|---|
| miRBase Accession Number: | MI0030344 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCUGCAGGCCCACUUCCCU |
| Species: | Mouse, Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0002093, mir-3552 |
Mature miRNA Sequence: | AGGCUGCAGGCCCACUUCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3552 | MIMAT0035715 |
| Rat | rno-miR-3552 | MIMAT0017813 |