Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn04229733_pri
Assay Name:
rno-mir-3552
Stem-loop ID:
rno-mir-3552
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.X: 118121374 - 118121478 [+] on Build Rnor_6.0
Stem-loop Location:
Chr.X: 118121374 - 118121478 [+] on Build Rnor_6.0
Mature miRNA ID:
rno-miR-3552
Species:
Rat
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Rn04229733_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2,900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
104
Chromosome Location:
Chr. X - 118121630 - 118121630 [+] on 118121630 Build Rnor_6.0
Species:
Rat
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Rat
rno-miR-3552
MIMAT0017813
Stem-loop Details
Rat
Stem-loop ID
rno-mir-3552
Stem-loop Accession #
MI0015409
Stem-loop Sequence
UGUGAAUCCAGCUGGUUCAAGGCUGCAGGCCCACUUCCCUUGGCAGAUUUUUCCAUUUCAGGGAGUGGGGAAAGUGGCCCUAAACCAUCAAAAUGGAUUAUACAU
Chromosome Location
Chr. X - 118121374 - 118121478 [+] on Build Rnor_6.0
Mature MicroRNA
miRBase Accession #: MIMAT0017813
miRBase ID: rno-miR-3552
Mature miRNA Sequence: AGGCUGCAGGCCCACUUCCCU
Chromosome Location: Chr. X - 118121374 - 118121478 [+] on Build Rnor_6.0
Gene Family ID
MIPF0002093,
mir-3552
Distance to Amplicon
106 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM20032
Ambion
®
Pre-miR
™
miRNA Precursor :
PM20032
TaqMan
™
MicroRNA Assay :
464903_mat
mir
Vana
®
miRNA inhibitor :
MH20032
mir
Vana
®
miRNA mimic :
MC20032
TaqMan
™
Advanced miRNA Assay :
rno481095_mir
TaqMan
™
Advanced miRNA Assay :
mmu482954_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.56.0-Offline