Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 465329_mat
          Assay Name: mmu-miR-669m-5p
          miRBase Accession Number: MI0014050
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 10466663 - 10466746 [+] on Build GRCm38
          Mature miRNA Sequence: UGUGUGCAUGUGCAUGUGUGUAU
          Species: Mouse
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 465329_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000316, mir-467

          Mature miRNA Sequence:

          UGUGUGCAUGUGCAUGUGUGUAU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-466m-5p MIMAT0014882
          Mouse mmu-miR-669m-5p MIMAT0017346

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-466m
          Stem-loop Accession # MI0014050
          Stem-loop Sequence
          UGUGUGUGUUCUUGUGUGUGCAUGUGCAUGUGUGUAUAUGAAUAUACAUAUACAUACACACAUACACACGCACGUACACACACA
          Chromosome Location Chr. 2 - 10466663 - 10466746 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014882
          miRBase ID: mmu-miR-466m-5p
          Mature miRNA Sequence: UGUGUGCAUGUGCAUGUGUGUAU
          Chromosome Location: Chr. 2 - 10466663 - 10466746 [+] on Build GRCm38

          Stem-loop ID mmu-mir-669m-1
          Stem-loop Accession # MI0009943
          Stem-loop Sequence
          UGUAUGUGCAUGUGUGUAUAGUUUUGUGUGCAUGUGCAUGUGUGUAUAUGAAUAGACAUAUACAUCCACACAAACAUAUACAAGCAAGCACAGAUACA
          Chromosome Location Chr. 2 - 10512790 - 10512887 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017346
          miRBase ID: mmu-miR-669m-5p
          Mature miRNA Sequence: UGUGUGCAUGUGCAUGUGUGUAU
          Chromosome Location: Chr. 2 - 10512790 - 10512887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-669m-2
          Stem-loop Accession # MI0009944
          Stem-loop Sequence
          AUAUUUGCAUGUGUGUAUAGUUUUGUGUGCAUGUGCAUGUGUGUAUAUGAAUAUACAUAUACAUCCACACAAACAUAUACAAGCAUGCACAGAUAUAC
          Chromosome Location Chr. 2 - 10513434 - 10513531 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017346
          miRBase ID: mmu-miR-669m-5p
          Mature miRNA Sequence: UGUGUGCAUGUGCAUGUGUGUAU
          Chromosome Location: Chr. 2 - 10512790 - 10512887 [+] on Build GRCm38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04228546_pri
          TaqMan™ Pri-miRNA Assay : Mm04227891_pri
          TaqMan™ Pri-miRNA Assay : Mm04227900_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19591
          Ambion® Pre-miR™ miRNA Precursor : PM19591
          mirVana® miRNA inhibitor : MH19591
          mirVana® miRNA mimic : MC19591
          TaqMan™ Advanced miRNA Assay : mmu483004_mir

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0