Search
Search

| Assay Name: | cgr-miR-671-5p |
|---|---|
| miRBase Accession Number: | MI0020549 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGAAGCCCUGGAGGGGCUGGAGG |
| Species: | Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Pig, Rabbit, Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000358, mir-671 |
Mature miRNA Sequence: | AGGAAGCCCUGGAGGGGCUGGAGG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Callithrix jacchus | cja-miR-671 | MIMAT0039371 | |
| Cricetulus griseus | cgr-miR-671-5p | MIMAT0023996 | |
| Dasypus novemcinctus | dno-miR-671-5p | MIMAT0047998 | |
| Goat | chi-miR-671-5p | MIMAT0036286 | |
| Guinea pig | cpo-miR-671-5p | MIMAT0047369 | |
| Pig | ssc-miR-671-5p | MIMAT0025381 | |
| View More Rows | |||
| Rabbit | ocu-miR-671-5p | MIMAT0048555 | |
| Tupaia chinensis | tch-miR-671-5p | MIMAT0036579 | |