Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11710
          Assay Name: cgr-miR-671-5p
          miRBase Accession Number: MI0020549
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Species: Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Pig, Rabbit, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC11710

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000358, mir-671

          Mature miRNA Sequence:

          AGGAAGCCCUGGAGGGGCUGGAGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Callithrix jacchus cja-miR-671 MIMAT0039371
          Cricetulus griseus cgr-miR-671-5p MIMAT0023996
          Dasypus novemcinctus dno-miR-671-5p MIMAT0047998
          Goat chi-miR-671-5p MIMAT0036286
          Guinea pig cpo-miR-671-5p MIMAT0047369
          Pig ssc-miR-671-5p MIMAT0025381
          View More Rows
          Rabbit ocu-miR-671-5p MIMAT0048555
          Tupaia chinensis tch-miR-671-5p MIMAT0036579

          Stem-loop Details



          Callithrix jacchus

          Stem-loop ID cja-mir-671
          Stem-loop Accession # MI0031889
          Stem-loop Sequence
          GGAAGAGGAGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACCUCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039371
          miRBase ID: cja-miR-671
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-671
          Stem-loop Accession # MI0020549
          Stem-loop Sequence
          CAGGAAGAGGAGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACCUCUUUCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023996
          miRBase ID: cgr-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-671
          Stem-loop Accession # MI0039166
          Stem-loop Sequence
          AGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047998
          miRBase ID: dno-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-671
          Stem-loop Accession # MI0030830
          Stem-loop Sequence
          GCCGCCGACCUGGCAGGCCAGGAAGAGGAGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACCUUUUCCGGGCCGUGGAGCCAGGGCUGGUGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036286
          miRBase ID: chi-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-671
          Stem-loop Accession # MI0038834
          Stem-loop Sequence
          AGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047369
          miRBase ID: cpo-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-671
          Stem-loop Accession # MI0022155
          Stem-loop Sequence
          CAGGAAGAGGAGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACCUUUUCCGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025381
          miRBase ID: ssc-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-671
          Stem-loop Accession # MI0039460
          Stem-loop Sequence
          AGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048555
          miRBase ID: ocu-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-671
          Stem-loop Accession # MI0031243
          Stem-loop Sequence
          AGGAAGCCCUGGAGGGGCUGGAGGUGAUGGAUGUUUUCCUCCGGUUCUCAGGGCUCCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036579
          miRBase ID: tch-miR-671-5p
          Mature miRNA Sequence: AGGAAGCCCUGGAGGGGCUGGAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11710
          Ambion® Pre-miR™ miRNA Precursor : PM11710
          TaqMan™ MicroRNA Assay : 465774_mat
          mirVana® miRNA inhibitor : MH11710

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline