Search
Search

| Assay Name: | bdi-miR528 |
|---|---|
| miRBase Accession Number: | MI0031699 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAGGGGCAUGCAGAGGAG |
| Species: | Aegilops tauschii, Brachypodium distachyon, Rice, Saccharum sp., Sorghum bicolor, Vriesea carinata, Zea mays |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000868, MIR528 |
Mature miRNA Sequence: | UGGAAGGGGCAUGCAGAGGAG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Aegilops tauschii | ata-miR528-5p | MIMAT0037192 | |
| Brachypodium distachyon | bdi-miR528-5p | MIMAT0020662 | |
| Rice | osa-miR528-5p | MIMAT0002884 | |
| Saccharum sp. | ssp-miR528 | MIMAT0020288 | |
| Sorghum bicolor | sbi-miR528 | MIMAT0014048 | |
| Vriesea carinata | vca-miR528-5p | MIMAT0041019 | |
| View More Rows | |||
| Zea mays | zma-miR528a-5p | MIMAT0014028 | |
| Zea mays | zma-miR528b-5p | MIMAT0014029 | |