Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10887
          Assay Name: ata-miR528-5p
          miRBase Accession Number: MI0031699
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Species: Aegilops tauschii, Brachypodium distachyon, Rice, Saccharum sp., Sorghum bicolor, Vriesea carinata, Zea mays
          Product Type: mirVana® miRNA mimic
          Assay ID MC10887

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000868, MIR528

          Mature miRNA Sequence:

          UGGAAGGGGCAUGCAGAGGAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Aegilops tauschii ata-miR528-5p MIMAT0037192
          Brachypodium distachyon bdi-miR528-5p MIMAT0020662
          Rice osa-miR528-5p MIMAT0002884
          Saccharum sp. ssp-miR528 MIMAT0020288
          Sorghum bicolor sbi-miR528 MIMAT0014048
          Vriesea carinata vca-miR528-5p MIMAT0041019
          View More Rows
          Zea mays zma-miR528a-5p MIMAT0014028
          Zea mays zma-miR528b-5p MIMAT0014029

          Stem-loop Details



          Aegilops tauschii

          Stem-loop ID ata-MIR528
          Stem-loop Accession # MI0031699
          Stem-loop Sequence
          GCAGCAGCGGUGGAAGGGGCAUGCAGAGGAGCGGCCAUGGCCCAUGGGAGCUUUGCUUGCCUUGCCUCUCGCUCUCUCCUGUGCCUGCCUCUUCCAUUCCUGCGGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037192
          miRBase ID: ata-miR528-5p
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Brachypodium distachyon

          Stem-loop ID bdi-MIR528
          Stem-loop Accession # MI0018087
          Stem-loop Sequence
          UUUGGGGUUGGGAUUGGGGGCUGGAGCAGCAGCGGUGGAAGGGGCAUGCAGAGGAGCAGGGAUGAUGGGGGGUUGUACUCUGCUUCUUCCCCUCUCGUUCUCCUGUGCCUGCCUCUUCCAUUCCUGCUGCUACCGCCGUGCUCGGUGGGCAGGGCUGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020662
          miRBase ID: bdi-miR528-5p
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Rice

          Stem-loop ID osa-MIR528
          Stem-loop Accession # MI0003201
          Stem-loop Sequence
          AGUGGAAGGGGCAUGCAGAGGAGCAGGAGAUUCAGUUUGAAGCUGGACUUCACUUUUGCCUCUCUCUCCUGUGCUUGCCUCUUCCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002884
          miRBase ID: osa-miR528-5p
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Saccharum sp.

          Stem-loop ID ssp-MIR528
          Stem-loop Accession # MI0018188
          Stem-loop Sequence
          UGGAAGGGGCAUGCAGAGGAGCACGAGGCUUGGCUAAAAGCCGACCUGUGCUAUGCUACCCCUCGCUGGCUCUCCUGUGCCUGCCUCUUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020288
          miRBase ID: ssp-miR528
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Sorghum bicolor

          Stem-loop ID sbi-MIR528
          Stem-loop Accession # MI0013259
          Stem-loop Sequence
          UGGAAGGGGCAUGCAGAGGAGCACGAGGCUUGGCUGGAAGCCGACCUACGCCUCGCUGGCUCUCCUGUGCCUGCCUCUUCCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014048
          miRBase ID: sbi-miR528
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Vriesea carinata

          Stem-loop ID vca-MIR528
          Stem-loop Accession # MI0032996
          Stem-loop Sequence
          GUGGAAGGGGCAUGCAGAGGAGAGGAGAGAUCCAAAACUCUACCCAGGCCCUGUUUUUCUUUUCCUGUGCUCGCCCCUUCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041019
          miRBase ID: vca-miR528-5p
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Zea mays

          Stem-loop ID zma-MIR528a
          Stem-loop Accession # MI0013239
          Stem-loop Sequence
          GUGGAAGGGGCAUGCAGAGGAGGAGCACGAGCGAGGUGUGGCUGGAAGAAGCAGCCGGGGCCUGUGUGCUAUAUACCCCUCGCAAGCUCUCCUCACUCCUCUCCUGUGCCUGCCUCUUCCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014028
          miRBase ID: zma-miR528a-5p
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA

          Stem-loop ID zma-MIR528b
          Stem-loop Accession # MI0013240
          Stem-loop Sequence
          GUGGAAGGGGCAUGCAGAGGAGCACGAACGAGGUGUGGUUGGCCCCUCGUUAGCUCUCCUGUGCCUGCCUCUUCCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014029
          miRBase ID: zma-miR528b-5p
          Mature miRNA Sequence: UGGAAGGGGCAUGCAGAGGAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 466226_mat
          mirVana® miRNA inhibitor : MH10887


          miRBase Alias:

          Brevundimonas diminuta: bdi-miR528 (v19)
          Oryza sativa: osa-miR528 (v18)
          Zea mays: zma-miR528a (v18)
          Zea mays: zma-miR528b (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline