Search
Search

| Assay Name: | hsa-miR-153-5p |
|---|---|
| miRBase Accession Number: | MI0000464 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAUUUUUGUGAUGUUGCAGCU |
| Species: | Human, Chicken, Monodelphis domestica, Ornithorhynchus anatinus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UCAUUUUUGUGAUGUUGCAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-153-5p | MIMAT0026480 |
| Chicken | gga-miR-153-5p | MIMAT0026502 |
| Monodelphis domestica | mdo-miR-153-2-5p | MIMAT0026931 |
| Ornithorhynchus anatinus | oan-miR-153-2-5p | MIMAT0031116 |
| Rhesus monkey | mml-miR-153-2-5p | MIMAT0031060 |