Search
Search

| Assay Name: | hsa-miR-653-3p |
|---|---|
| miRBase Accession Number: | MI0003674 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCACUGGAGUUUGUUUCAAUA |
| Species: | Human, Cricetulus griseus, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000435, mir-653 |
Mature miRNA Sequence: | UUCACUGGAGUUUGUUUCAAUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-653-3p | MIMAT0026625 |
| Cricetulus griseus | cgr-miR-653 | MIMAT0023993 |
| Rabbit | ocu-miR-653-3p | MIMAT0048544 |
| Rhesus monkey | mml-miR-653-3p | MIMAT0026905 |