Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477861_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-let-7a-3p
          Stem-loop Accession Number: MI0000060
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 94175957 - 94176036 [+] on Build GRCh38
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Species: Human, Anolis carolinensis, Bovine, Columba livia, Cricetulus griseus
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477861_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUAUACAAUCUACUGUCUUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7a-3p MIMAT0004481
          Anolis carolinensis aca-let-7a-3-3p MIMAT0037558
          Bovine bta-let-7a-3p MIMAT0004330
          Columba livia cli-let-7a-3p MIMAT0038375
          Cricetulus griseus cgr-let-7c-2 MIMAT0041131

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7a-1
          Stem-loop Accession # MI0000060
          Stem-loop Sequence
          UGGGAUGAGGUAGUAGGUUGUAUAGUUUUAGGGUCACACCCACCACUGGGAGAUAACUAUACAAUCUACUGUCUUUCCUA
          Chromosome Location Chr. 9 - 94175957 - 94176036 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004481
          miRBase ID: hsa-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: Chr. 22 - 46112749 - 46112822 [+] on Build GRCh38

          Stem-loop ID hsa-let-7a-3
          Stem-loop Accession # MI0000062
          Stem-loop Sequence
          GGGUGAGGUAGUAGGUUGUAUAGUUUGGGGCUCUGCCCUGCUAUGGGAUAACUAUACAAUCUACUGUCUUUCCU
          Chromosome Location Chr. 22 - 46112749 - 46112822 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004481
          miRBase ID: hsa-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: Chr. 22 - 46112749 - 46112822 [+] on Build GRCh38

          Anolis carolinensis

          Stem-loop ID aca-let-7a-3
          Stem-loop Accession # MI0029347
          Stem-loop Sequence
          GGGUGAGGUAGUAGGUUGUAUAGUUUUAGGGUUCUGCCCUGCCUGUCACAUAACUAUACAAUCUACUGUCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037558
          miRBase ID: aca-let-7a-3-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-let-7a-3
          Stem-loop Accession # MI0005452
          Stem-loop Sequence
          GGGUGAGGUAGUAGGUUGUAUAGUUUGGGGCUCUGCCCUGCUAUGGGAUAACUAUACAAUCUACUGUCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004330
          miRBase ID: bta-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7a-1
          Stem-loop Accession # MI0029882
          Stem-loop Sequence
          CUGUGCUGUGGGAUGAGGUAGUAGGUUGUAUAGUUUUAGGGUCAUACCCGCAACUGGGAGAUAACUAUACAAUCUACUGUCUUUCCUAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038375
          miRBase ID: cli-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: NA

          Stem-loop ID cli-let-7a-3
          Stem-loop Accession # MI0029884
          Stem-loop Sequence
          GUCCUUUGGGGUGAGGUAGUAGGUUGUAUAGUUUUAGGGCUCUGCCCUGCCUGUCACAUAACUAUACAAUCUACUGUCUUUCCUGAAGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038375
          miRBase ID: cli-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7c-2
          Stem-loop Accession # MI0033059
          Stem-loop Sequence
          GGGUGAGGUAGUAGGUUGUAUGGUUUUGGGCUCUGCCCCGCUCUGCGGUAACUAUACAAUCUACUGUCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041131
          miRBase ID: cgr-let-7c-2
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302546_pri
          TaqMan™ Pri-miRNA Assay : Hs03302533_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12169
          Ambion® Pre-miR™ miRNA Precursor : PM12169
          TaqMan™ MicroRNA Assay : 002307
          mirVana® miRNA inhibitor : MH12169
          mirVana® miRNA mimic : MC12169
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 3
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 3
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch