Search
Search

| Assay Name: | hsa-miR-105-5p |
|---|---|
| Stem-loop Accession Number: | MI0000111 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUGUGGU |
| Species: | Human, Bovine, Horse, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUGUGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-105-5p | MIMAT0000102 |
| Bovine | bta-miR-105a | MIMAT0009217 |
| Horse | eca-miR-105 | MIMAT0013193 |
| Pongo pygmaeus | ppy-miR-105 | MIMAT0002450 |
| Rhesus monkey | mml-miR-105-5p | MIMAT0002454 |