Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12838
          Assay Name: hsa-miR-105-5p
          miRBase Accession Number: MI0000111
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 152392219 - 152392299 [-] on Build GRCh38
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Species: Human, Bovine, Horse, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12838

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000074, mir-105

          Mature miRNA Sequence:

          UCAAAUGCUCAGACUCCUGUGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-105-5p MIMAT0000102
          Bovine bta-miR-105a MIMAT0009217
          Horse eca-miR-105 MIMAT0013193
          Pongo pygmaeus ppy-miR-105 MIMAT0002450
          Rhesus monkey mml-miR-105-5p MIMAT0002454

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-105-1
          Stem-loop Accession # MI0000111
          Stem-loop Sequence
          UGUGCAUCGUGGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUCAUGCACCACGGAUGUUUGAGCAUGUGCUACGGUGUCUA
          Chromosome Location Chr. X - 152392219 - 152392299 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000102
          miRBase ID: hsa-miR-105-5p
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: Chr. X - 152394412 - 152394492 [-] on Build GRCh38

          Stem-loop ID hsa-mir-105-2
          Stem-loop Accession # MI0000112
          Stem-loop Sequence
          UGUGCAUCGUGGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUUAUGCACCACGGAUGUUUGAGCAUGUGCUAUGGUGUCUA
          Chromosome Location Chr. X - 152394412 - 152394492 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000102
          miRBase ID: hsa-miR-105-5p
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: Chr. X - 152394412 - 152394492 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-105a
          Stem-loop Accession # MI0009723
          Stem-loop Sequence
          UGUGCAUCGUAGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUCAUGCACCACGGAUGUUUGAGCAUGUGCUACGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009217
          miRBase ID: bta-miR-105a
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-105
          Stem-loop Accession # MI0012941
          Stem-loop Sequence
          UGUGCAUCGUAGUCAAAUGCUCAGACUCCUGUGGUGGCCGUUCAUGCACCACGGAUGUUUGAGCAUGUGCUAUGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013193
          miRBase ID: eca-miR-105
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-105-1
          Stem-loop Accession # MI0002746
          Stem-loop Sequence
          UGUGCAUCGUGGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUCAUGCACCACGGAUGUUUGAGCAUGUGCUACGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002450
          miRBase ID: ppy-miR-105
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: NA

          Stem-loop ID ppy-mir-105-2
          Stem-loop Accession # MI0014814
          Stem-loop Sequence
          UGUGCAUCGUGGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUUAUGCACCACGGAUGUUUGAGCAUGUGCUAUGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002450
          miRBase ID: ppy-miR-105
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-105-1
          Stem-loop Accession # MI0002750
          Stem-loop Sequence
          UGUGCAUCGUGGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUCAUGCACCACGGAUGUUUGAGCAUGUGCUACGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002454
          miRBase ID: mml-miR-105-5p
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: NA

          Stem-loop ID mml-mir-105-2
          Stem-loop Accession # MI0007612
          Stem-loop Sequence
          UGUGCAUCGUGGUCAAAUGCUCAGACUCCUGUGGUGGCUGCUUAUGCACCACGGAUGUUUGAGCAUGUGCUAUGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002454
          miRBase ID: mml-miR-105-5p
          Mature miRNA Sequence: UCAAAUGCUCAGACUCCUGUGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302772_pri
          TaqMan™ Pri-miRNA Assay : Hs03302770_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12838
          Ambion® Pre-miR™ miRNA Precursor : PM12838
          TaqMan™ MicroRNA Assay : 002167
          mirVana® miRNA inhibitor : MH12838
          TaqMan™ Advanced miRNA Assay : 477865_mir


          miRBase Alias:

          Human: hsa-miR-105 (v17)
          Macaca mulatta: mml-miR-105 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline