Search
Search

| Assay Name: | hsa-miR-326 |
|---|---|
| Stem-loop Accession Number: | MI0000808 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUCUGGGCCCUUCCUCCAG |
| Species: | Human, Bovine, Dasypus novemcinctus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000143, mir-326 |
Mature miRNA Sequence: | CCUCUGGGCCCUUCCUCCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-326 | MIMAT0000756 |
| Bovine | bta-miR-326 | MIMAT0009286 |
| Dasypus novemcinctus | dno-miR-326-3p | MIMAT0047806 |
| Dog | cfa-miR-326 | MIMAT0009894 |
| Gorilla gorilla | ggo-miR-326 | MIMAT0024215 |
| Guinea pig | cpo-miR-326-3p | MIMAT0047161 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-326 | MIMAT0008099 |
| Pig | ssc-miR-326 | MIMAT0002140 |
| Pongo pygmaeus | ppy-miR-326 | MIMAT0015830 |
| Rabbit | ocu-miR-326-3p | MIMAT0048375 |