Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10686
          Assay Name: hsa-miR-326
          miRBase Accession Number: MI0000808
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 75335092 - 75335186 [-] on Build GRCh38
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Species: Human, Bovine, Dasypus novemcinctus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10686

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000143, mir-326

          Mature miRNA Sequence:

          CCUCUGGGCCCUUCCUCCAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-326 MIMAT0000756
          Bovine bta-miR-326 MIMAT0009286
          Dasypus novemcinctus dno-miR-326-3p MIMAT0047806
          Dog cfa-miR-326 MIMAT0009894
          Gorilla gorilla ggo-miR-326 MIMAT0024215
          Guinea pig cpo-miR-326-3p MIMAT0047161
          View More Rows
          Pan troglodytes ptr-miR-326 MIMAT0008099
          Pig ssc-miR-326 MIMAT0002140
          Pongo pygmaeus ppy-miR-326 MIMAT0015830
          Rabbit ocu-miR-326-3p MIMAT0048375

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-326
          Stem-loop Accession # MI0000808
          Stem-loop Sequence
          CUCAUCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGAAGGCGGGUGGUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGCCCCGAGGCGGAUUCA
          Chromosome Location Chr. 11 - 75335092 - 75335186 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000756
          miRBase ID: hsa-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: Chr. 11 - 75335092 - 75335186 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-326
          Stem-loop Accession # MI0009799
          Stem-loop Sequence
          CUCGUCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGAAGGCGGGUUGUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGCCCAGAGGCGGAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009286
          miRBase ID: bta-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-326
          Stem-loop Accession # MI0039067
          Stem-loop Sequence
          GGAGGCAGGGCCUUUGUGAAGGCGGGUUGUGCUCGGAUCGCCUCUGGGCCCUUCCUCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047806
          miRBase ID: dno-miR-326-3p
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-326
          Stem-loop Accession # MI0010393
          Stem-loop Sequence
          UCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGAAGGCGGGUGGUGAUCGGAUCGCCUCUGGGCCCUUCCUCCAGCCCAGAGGCGGAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009894
          miRBase ID: cfa-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-326
          Stem-loop Accession # MI0020762
          Stem-loop Sequence
          CUUCUGGAGCCUCAUCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGAAGGCGGGUGGUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGCCCCGAGGCGGAUUCACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024215
          miRBase ID: ggo-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-326
          Stem-loop Accession # MI0038728
          Stem-loop Sequence
          GGGGGCAGGGCCUUUGUGAAGGCGGGUGUGCAAAGAUCGCCUCUGGGCCCUUCCUCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047161
          miRBase ID: cpo-miR-326-3p
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-326
          Stem-loop Accession # MI0008617
          Stem-loop Sequence
          CUCAUCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGAAGGCGGGUGGUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGCCCCGAGGCGGAUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008099
          miRBase ID: ptr-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-326
          Stem-loop Accession # MI0002434
          Stem-loop Sequence
          UCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGCAGGCGGGUUGUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGCCCAGAGGCGGAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002140
          miRBase ID: ssc-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-326
          Stem-loop Accession # MI0014894
          Stem-loop Sequence
          CUCAUCUGUCUGUUGGGCUGGAGGCAGGGCCUUUGUGAAGGCGGGUGGUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGCCCCGAGGCGGAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015830
          miRBase ID: ppy-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-326
          Stem-loop Accession # MI0039367
          Stem-loop Sequence
          GGGGGCAGGGCCUUUGUGAAGGCGGGUUGUGCUCGGAUCGCCUCUGGGCCCUUCCUCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048375
          miRBase ID: ocu-miR-326-3p
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303601_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10686
          Ambion® Pre-miR™ miRNA Precursor : PM10686
          TaqMan™ MicroRNA Assay : 000542
          mirVana® miRNA mimic : MC10686
          TaqMan™ Advanced miRNA Assay : 478027_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline