Search
Search

| Assay Name: | hsa-miR-330-3p |
|---|---|
| Stem-loop Accession Number: | MI0000803 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCAAAGCACACGGCCUGCAGAGA |
| Species: | Human, Bovine, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000200, mir-330 |
Mature miRNA Sequence: | GCAAAGCACACGGCCUGCAGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-330-3p | MIMAT0000751 |
| Bovine | bta-miR-330 | MIMAT0009290 |
| Gorilla gorilla | ggo-miR-330 | MIMAT0024088 |
| Pan troglodytes | ptr-miR-330 | MIMAT0008102 |
| Pongo pygmaeus | ppy-miR-330-3p | MIMAT0015834 |
| Pteropus alecto | pal-miR-330-3p | MIMAT0040096 |
| View More Rows | ||
| Rhesus monkey | mml-miR-330-3p | MIMAT0006271 |
| Tupaia chinensis | tch-miR-330-3p | MIMAT0036574 |