Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10735
          Assay Name: hsa-miR-330-3p
          miRBase Accession Number: MI0000803
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 45638994 - 45639087 [-] on Build GRCh38
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Species: Human, Bovine, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10735
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000200, mir-330

          Mature miRNA Sequence:

          GCAAAGCACACGGCCUGCAGAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-330-3p MIMAT0000751
          Bovine bta-miR-330 MIMAT0009290
          Gorilla gorilla ggo-miR-330 MIMAT0024088
          Pan troglodytes ptr-miR-330 MIMAT0008102
          Pongo pygmaeus ppy-miR-330-3p MIMAT0015834
          Pteropus alecto pal-miR-330-3p MIMAT0040096
          View More Rows
          Rhesus monkey mml-miR-330-3p MIMAT0006271
          Tupaia chinensis tch-miR-330-3p MIMAT0036574

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-330
          Stem-loop Accession # MI0000803
          Stem-loop Sequence
          CUUUGGCGAUCACUGCCUCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGAGGCAGCGCUCUGCCC
          Chromosome Location Chr. 19 - 45638994 - 45639087 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000751
          miRBase ID: hsa-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: Chr. 19 - 45638994 - 45639087 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-330
          Stem-loop Accession # MI0009803
          Stem-loop Sequence
          CUUCGGCGAUCACUGCCUCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGAGGCAGCGCUCAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009290
          miRBase ID: bta-miR-330
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-330
          Stem-loop Accession # MI0020635
          Stem-loop Sequence
          UCGUGCUGUGUGAUCUUUGGCGAUCACUGCCUCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGAGGCAGCGCUCUGCCCCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024088
          miRBase ID: ggo-miR-330
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-330
          Stem-loop Accession # MI0008621
          Stem-loop Sequence
          CUUUGGCGAUCACUGCCUCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGAGGCAGCGCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008102
          miRBase ID: ptr-miR-330
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-330
          Stem-loop Accession # MI0014898
          Stem-loop Sequence
          CUUUGGCGAUCACUGCCUCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGAGGCAGCGCUCUGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015834
          miRBase ID: ppy-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-330
          Stem-loop Accession # MI0032491
          Stem-loop Sequence
          UCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUGAACCGAGCAAAGCACACGGCCUGCAGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040096
          miRBase ID: pal-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-330
          Stem-loop Accession # MI0007697
          Stem-loop Sequence
          CUUUGGCGAUCACUGCCUCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGAGGCAGCGCUCUGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006271
          miRBase ID: mml-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-330
          Stem-loop Accession # MI0031237
          Stem-loop Sequence
          UCUCUGGGCCUGUGUCUUAGGCUCUGCAAGAUCAACCGAGCAAAGCACACGGCCUGCAGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036574
          miRBase ID: tch-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACACGGCCUGCAGAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303577_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10735
          TaqMan™ MicroRNA Assay : 000544
          mirVana® miRNA inhibitor : MH10735
          mirVana® miRNA mimic : MC10735
          TaqMan™ Advanced miRNA Assay : 478030_mir


          miRBase Alias:

          Human: hsa-miR-330 (v9.2)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline