Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478105_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-448
          Stem-loop Accession Number: MI0001637
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 114823454 - 114823564 [+] on Build GRCh38
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Species: Human, Mouse, Bovine, Dog, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478105_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000149, mir-448

          Mature miRNA Sequence:

          UUGCAUAUGUAGGAUGUCCCAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-448 MIMAT0001532
          Mouse mmu-miR-448-3p MIMAT0001533
          Bovine bta-miR-448 MIMAT0009319
          Dog cfa-miR-448 MIMAT0001535
          Horse eca-miR-448 MIMAT0013218
          Pan troglodytes ptr-miR-448 MIMAT0008148
          View More Rows
          Pongo pygmaeus ppy-miR-448 MIMAT0015890
          Rhesus monkey mml-miR-448 MIMAT0006329

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-448
          Stem-loop Accession # MI0001637
          Stem-loop Sequence
          GCCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUAUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCCACUUCGUCA
          Chromosome Location Chr. X - 114823454 - 114823564 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001532
          miRBase ID: hsa-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: Chr. X - 114823454 - 114823564 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-448
          Stem-loop Accession # MI0001638
          Stem-loop Sequence
          ACGAGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUACUUCAUACUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCUGGCCCACUUCGUCA
          Chromosome Location Chr. X - 147158210 - 147158321 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001533
          miRBase ID: mmu-miR-448-3p
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: Chr. X - 147158210 - 147158321 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-448
          Stem-loop Accession # MI0009833
          Stem-loop Sequence
          GCGGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUUAUACUAAGGGGGGGUUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCCACCCACAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009319
          miRBase ID: bta-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-448
          Stem-loop Accession # MI0001640
          Stem-loop Sequence
          GCAGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUUAUACUAAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCCGCCCACUUCAUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001535
          miRBase ID: cfa-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-448
          Stem-loop Accession # MI0012964
          Stem-loop Sequence
          GGCGGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCCAUUUUAUACUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCCGCCCGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013218
          miRBase ID: eca-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-448
          Stem-loop Accession # MI0008669
          Stem-loop Sequence
          CCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUAUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCCACUUCGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008148
          miRBase ID: ptr-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-448
          Stem-loop Accession # MI0014946
          Stem-loop Sequence
          GCCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUAUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCCACUUCGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015890
          miRBase ID: ppy-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-448
          Stem-loop Accession # MI0007746
          Stem-loop Sequence
          GCCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUACUAAGAGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCUACUUCGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006329
          miRBase ID: mml-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307489_pri
          TaqMan™ Pri-miRNA Assay : Hs03303721_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10520
          Ambion® Pre-miR™ miRNA Precursor : PM10520
          TaqMan™ MicroRNA Assay : 001029
          mirVana® miRNA inhibitor : MH10520
          mirVana® miRNA mimic : MC10520
          TaqMan™ Advanced miRNA Assay : mmu481839_mir
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 4
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 4
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A Card

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline