Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10520
          Assay Name: hsa-miR-448
          miRBase Accession Number: MI0001637
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 114823454 - 114823564 [+] on Build GRCh38
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Species: Human, Mouse, Bovine, Dog, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC10520

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000149, mir-448

          Mature miRNA Sequence:

          UUGCAUAUGUAGGAUGUCCCAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-448 MIMAT0001532
          Mouse mmu-miR-448-3p MIMAT0001533
          Bovine bta-miR-448 MIMAT0009319
          Dog cfa-miR-448 MIMAT0001535
          Horse eca-miR-448 MIMAT0013218
          Pan troglodytes ptr-miR-448 MIMAT0008148
          View More Rows
          Pongo pygmaeus ppy-miR-448 MIMAT0015890
          Rhesus monkey mml-miR-448 MIMAT0006329

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-448
          Stem-loop Accession # MI0001637
          Stem-loop Sequence
          GCCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUAUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCCACUUCGUCA
          Chromosome Location Chr. X - 114823454 - 114823564 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001532
          miRBase ID: hsa-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: Chr. X - 114823454 - 114823564 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-448
          Stem-loop Accession # MI0001638
          Stem-loop Sequence
          ACGAGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUACUUCAUACUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCUGGCCCACUUCGUCA
          Chromosome Location Chr. X - 147158210 - 147158321 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001533
          miRBase ID: mmu-miR-448-3p
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: Chr. X - 147158210 - 147158321 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-448
          Stem-loop Accession # MI0009833
          Stem-loop Sequence
          GCGGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUUAUACUAAGGGGGGGUUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCCACCCACAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009319
          miRBase ID: bta-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-448
          Stem-loop Accession # MI0001640
          Stem-loop Sequence
          GCAGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUUAUACUAAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCCGCCCACUUCAUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001535
          miRBase ID: cfa-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-448
          Stem-loop Accession # MI0012964
          Stem-loop Sequence
          GGCGGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCCAUUUUAUACUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCCGCCCGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013218
          miRBase ID: eca-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-448
          Stem-loop Accession # MI0008669
          Stem-loop Sequence
          CCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUAUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCCACUUCGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008148
          miRBase ID: ptr-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-448
          Stem-loop Accession # MI0014946
          Stem-loop Sequence
          GCCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUAUAAGAGGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCCACUUCGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015890
          miRBase ID: ppy-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-448
          Stem-loop Accession # MI0007746
          Stem-loop Sequence
          GCCGGGAGGUUGAACAUCCUGCAUAGUGCUGCCAGGAAAUCCCUAUUUCAUACUAAGAGGGGCUGGCUGGUUGCAUAUGUAGGAUGUCCCAUCUCCCAGCCUACUUCGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006329
          miRBase ID: mml-miR-448
          Mature miRNA Sequence: UUGCAUAUGUAGGAUGUCCCAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307489_pri
          TaqMan™ Pri-miRNA Assay : Hs03303721_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10520
          Ambion® Pre-miR™ miRNA Precursor : PM10520
          TaqMan™ MicroRNA Assay : 001029
          mirVana® miRNA inhibitor : MH10520
          TaqMan™ Advanced miRNA Assay : 478105_mir
          TaqMan™ Advanced miRNA Assay : mmu481839_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline