Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478107_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-451a
          Stem-loop Accession Number: MI0001729
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 28861369 - 28861440 [-] on Build GRCh38
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Species: Human, Mouse, Rat, Anolis carolinensis, Cricetulus griseus, Dog, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478107_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000148, mir-451

          Mature miRNA Sequence:

          AAACCGUUACCAUUACUGAGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-451a MIMAT0001631
          Mouse mmu-miR-451a MIMAT0001632
          Rat rno-miR-451-5p MIMAT0001633
          Anolis carolinensis aca-miR-451-5p MIMAT0021960
          Cricetulus griseus cgr-miR-451a MIMAT0041141
          Dog cfa-miR-451 MIMAT0009870
          View More Rows
          Pan troglodytes ptr-miR-451 MIMAT0008153
          Pig ssc-miR-451 MIMAT0018382
          Pongo pygmaeus ppy-miR-451 MIMAT0015896
          Rhesus monkey mml-miR-451 MIMAT0006335
          Xenopus tropicalis xtr-miR-451 MIMAT0003705
          Zebrafish dre-miR-451 MIMAT0001634

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-451a
          Stem-loop Accession # MI0001729
          Stem-loop Sequence
          CUUGGGAAUGGCAAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAUGGUUCUCUUGCUAUACCCAGA
          Chromosome Location Chr. 17 - 28861369 - 28861440 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001631
          miRBase ID: hsa-miR-451a
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: Chr. 17 - 28861369 - 28861440 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-451a
          Stem-loop Accession # MI0001730
          Stem-loop Sequence
          CUUGGGAAUGGCGAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAACGGUUCUCUUGCUGCUCCCACA
          Chromosome Location Chr. 11 - 78073170 - 78073241 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001632
          miRBase ID: mmu-miR-451a
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: Chr. 11 - 78073170 - 78073241 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-451
          Stem-loop Accession # MI0001731
          Stem-loop Sequence
          UUUGGGAAUGGCGAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAUGGUUCUCUUGCUGCUCCCACA
          Chromosome Location Chr. 10 - 65291204 - 65291275 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001633
          miRBase ID: rno-miR-451-5p
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: Chr. 10 - 65291204 - 65291275 [-] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-451
          Stem-loop Accession # MI0018869
          Stem-loop Sequence
          UGACCGGUGUGUUGGCGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAGGGUUCUUCUAUCACGCCGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021960
          miRBase ID: aca-miR-451-5p
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-451a
          Stem-loop Accession # MI0033070
          Stem-loop Sequence
          CUUGGGGAUGGCGAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAACGGUUCUCUUGCUGCUCCCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041141
          miRBase ID: cgr-miR-451a
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-451
          Stem-loop Accession # MI0010367
          Stem-loop Sequence
          UUGGGGAUGGCGAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAUGGUUCUCUUGCUGUACCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009870
          miRBase ID: cfa-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-451
          Stem-loop Accession # MI0008675
          Stem-loop Sequence
          UUGGGAAUGGCAAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAUGGUUCUCUUGCUAUACCCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008153
          miRBase ID: ptr-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-451
          Stem-loop Accession # MI0016623
          Stem-loop Sequence
          GGGGGUGGCGAGCAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAACGGUUCUCUUGCUGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018382
          miRBase ID: ssc-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-451
          Stem-loop Accession # MI0014952
          Stem-loop Sequence
          CUUGGGAAUGGCAAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAUGGUUCUCUUGCUAUACCCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015896
          miRBase ID: ppy-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-451
          Stem-loop Accession # MI0007752
          Stem-loop Sequence
          CUUGGGAAUGGCAAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAGGGUUCUCUUGCUAUAUCCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006335
          miRBase ID: mml-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-451
          Stem-loop Accession # MI0004958
          Stem-loop Sequence
          GAGUGGCAAUGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAGGGUUCUGUUGCUGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003705
          miRBase ID: xtr-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-451
          Stem-loop Accession # MI0001732
          Stem-loop Sequence
          AGAGGCGGCGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAAGGGUUCUGCUGCCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001634
          miRBase ID: dre-miR-451
          Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295288_pri
          TaqMan™ Pri-miRNA Assay : Rn03465141_pri
          TaqMan™ Pri-miRNA Assay : Mm03307524_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10286
          Ambion® Pre-miR™ miRNA Precursor : PM10286
          TaqMan™ MicroRNA Assay : 001141
          mirVana® miRNA inhibitor : MH10286
          mirVana® miRNA mimic : MC10286
          TaqMan™ Advanced miRNA Assay : rno481395_mir
          TaqMan™ Advanced miRNA Assay : mmu482766_mir
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-8frr5:80/100.66.74.151:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline