Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03307524_pri
Assay Name:
mmu-mir-451a
Stem-loop ID:
mmu-mir-451a
mmu-mir-451b
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.11: 78073170 - 78073241 [+] on Build GRCm38
Stem-loop Location:
Chr.11: 78073170 - 78073241 [+] on Build GRCm38
Chr.11: 78073170 - 78073241 [-] on Build GRCm38
Mature miRNA ID:
mmu-miR-451a
mmu-miR-451b
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03307524_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2,900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
74
Chromosome Location:
Chr. 11 - 78073372 - 78073372 [-] on 78073372 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-451a
MIMAT0001632
Mouse
mmu-miR-451b
MIMAT0025178
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-451a
Stem-loop Accession #
MI0001730
Stem-loop Sequence
CUUGGGAAUGGCGAGGAAACCGUUACCAUUACUGAGUUUAGUAAUGGUAACGGUUCUCUUGCUGCUCCCACA
Chromosome Location
Chr. 11 - 78073170 - 78073241 [+] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0001632
miRBase ID: mmu-miR-451a
Mature miRNA Sequence: AAACCGUUACCAUUACUGAGUU
Chromosome Location: Chr. 11 - 78073170 - 78073241 [+] on Build GRCm38
Gene Family ID
MIPF0000148,
mir-451
Distance to Amplicon
102 bases
Stem-loop ID
mmu-mir-451b
Stem-loop Accession #
MI0021960
Stem-loop Sequence
UGUGGGAGCAGCAAGAGAACCGUUACCAUUACUAAACUCAGUAAUGGUAACGGUUUCCUCGCCAUUCCCAAG
Chromosome Location
Chr. 11 - 78073170 - 78073241 [-] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0025178
miRBase ID: mmu-miR-451b
Mature miRNA Sequence: UGGGAGCAGCAAGAGAACCGU
Chromosome Location: Chr. 11 - 78073170 - 78073241 [-] on Build GRCm38
Gene Family ID
MIPF0000148,
mir-451
Distance to Amplicon
102 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM24546
Ambion
®
Pre-miR
™
miRNA Precursor :
PM24546
TaqMan
™
MicroRNA Assay :
472639_mat
mir
Vana
®
miRNA inhibitor :
MH24546
mir
Vana
®
miRNA mimic :
MC24546
TaqMan
™
Advanced miRNA Assay :
mmu481843_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10286
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10286
TaqMan
™
MicroRNA Assay :
001141
mir
Vana
®
miRNA inhibitor :
MH10286
mir
Vana
®
miRNA mimic :
MC10286
TaqMan
™
Advanced miRNA Assay :
rno481395_mir
TaqMan
™
Advanced miRNA Assay :
mmu482766_mir
TaqMan
™
Advanced miRNA Assay :
478107_mir
miRBase Alias:
Mouse: mmu-mir-451a (v21)
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.57.0-Offline