Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478134_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-493-3p
          Stem-loop Accession Number: MI0003132
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100869060 - 100869148 [+] on Build GRCh38
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Species: Human, Bovine, Goat, Pig, Pongo pygmaeus, Rhesus monkey, Sheep
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478134_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000230, mir-493

          Mature miRNA Sequence:

          UGAAGGUCUACUGUGUGCCAGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-493-3p MIMAT0003161
          Bovine bta-miR-493 MIMAT0009333
          Goat chi-miR-493-3p MIMAT0036254
          Pig ssc-miR-493-3p MIMAT0025378
          Pongo pygmaeus ppy-miR-493 MIMAT0015914
          Rhesus monkey mml-miR-493-3p MIMAT0006355
          View More Rows
          Sheep oar-miR-493-3p MIMAT0019238

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-493
          Stem-loop Accession # MI0003132
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location Chr. 14 - 100869060 - 100869148 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003161
          miRBase ID: hsa-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: Chr. 14 - 100869060 - 100869148 [+] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-493
          Stem-loop Accession # MI0009847
          Stem-loop Sequence
          CUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009333
          miRBase ID: bta-miR-493
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-493
          Stem-loop Accession # MI0030807
          Stem-loop Sequence
          GGGGCUCCCUCUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGCAGCAAGUAAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036254
          miRBase ID: chi-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-493
          Stem-loop Accession # MI0022152
          Stem-loop Sequence
          CCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUGCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025378
          miRBase ID: ssc-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-493
          Stem-loop Accession # MI0014966
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015914
          miRBase ID: ppy-miR-493
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-493
          Stem-loop Accession # MI0007767
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006355
          miRBase ID: mml-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-493
          Stem-loop Accession # MI0016912
          Stem-loop Sequence
          GGGGCUCCCUCUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGCAGCAAGUAAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019238
          miRBase ID: oar-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293825_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12761
          Ambion® Pre-miR™ miRNA Precursor : PM12761
          TaqMan™ MicroRNA Assay : 002364
          mirVana® miRNA inhibitor : MH12761
          mirVana® miRNA mimic : MC12761
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline