Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12761
          Assay Name: hsa-miR-493-3p
          miRBase Accession Number: MI0003132
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100869060 - 100869148 [+] on Build GRCh38
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Species: Human, Bovine, Goat, Pig, Pongo pygmaeus, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA mimic
          Assay ID MC12761

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000230, mir-493

          Mature miRNA Sequence:

          UGAAGGUCUACUGUGUGCCAGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-493-3p MIMAT0003161
          Bovine bta-miR-493 MIMAT0009333
          Goat chi-miR-493-3p MIMAT0036254
          Pig ssc-miR-493-3p MIMAT0025378
          Pongo pygmaeus ppy-miR-493 MIMAT0015914
          Rhesus monkey mml-miR-493-3p MIMAT0006355
          View More Rows
          Sheep oar-miR-493-3p MIMAT0019238

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-493
          Stem-loop Accession # MI0003132
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location Chr. 14 - 100869060 - 100869148 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003161
          miRBase ID: hsa-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: Chr. 14 - 100869060 - 100869148 [+] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-493
          Stem-loop Accession # MI0009847
          Stem-loop Sequence
          CUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009333
          miRBase ID: bta-miR-493
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-493
          Stem-loop Accession # MI0030807
          Stem-loop Sequence
          GGGGCUCCCUCUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGCAGCAAGUAAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036254
          miRBase ID: chi-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-493
          Stem-loop Accession # MI0022152
          Stem-loop Sequence
          CCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUGCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025378
          miRBase ID: ssc-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-493
          Stem-loop Accession # MI0014966
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015914
          miRBase ID: ppy-miR-493
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-493
          Stem-loop Accession # MI0007767
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006355
          miRBase ID: mml-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-493
          Stem-loop Accession # MI0016912
          Stem-loop Sequence
          GGGGCUCCCUCUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGCAGCAAGUAAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019238
          miRBase ID: oar-miR-493-3p
          Mature miRNA Sequence: UGAAGGUCUACUGUGUGCCAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293825_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12761
          Ambion® Pre-miR™ miRNA Precursor : PM12761
          TaqMan™ MicroRNA Assay : 002364
          mirVana® miRNA inhibitor : MH12761
          TaqMan™ Advanced miRNA Assay : 478134_mir


          miRBase Alias:

          Human: hsa-miR-493 (v17)
          Macaca mulatta: mml-miR-493 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline