Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478158_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-551a
          Stem-loop Accession Number: MI0003556
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 3560695 - 3560790 [-] on Build GRCh38
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Species: Human, Dog, Gorilla gorilla, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478158_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000360, mir-551

          Mature miRNA Sequence:

          GCGACCCACUCUUGGUUUCCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-551a MIMAT0003214
          Dog cfa-miR-551a MIMAT0009912
          Gorilla gorilla ggo-miR-551a MIMAT0024186
          Horse eca-miR-551a MIMAT0012921
          Pan troglodytes ptr-miR-551a MIMAT0008231
          Pongo pygmaeus ppy-miR-551a MIMAT0015997
          View More Rows
          Rhesus monkey mml-miR-551a MIMAT0006419

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-551a
          Stem-loop Accession # MI0003556
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCAGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAA
          Chromosome Location Chr. 1 - 3560695 - 3560790 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003214
          miRBase ID: hsa-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: Chr. 1 - 3560695 - 3560790 [-] on Build GRCh38

          Dog

          Stem-loop ID cfa-mir-551a
          Stem-loop Accession # MI0010411
          Stem-loop Sequence
          GGGGACUGCCAGGUGACCCUGGAAAUCCAGAGUGGGUAAGGCCUGUCUGAUUGUGUCUAGGCGACCCACUCUUGGUUUCCAUGGUUGCCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009912
          miRBase ID: cfa-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-551a
          Stem-loop Accession # MI0020733
          Stem-loop Sequence
          GGCUGCCAGAGGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAACCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024186
          miRBase ID: ggo-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-551a
          Stem-loop Accession # MI0012677
          Stem-loop Sequence
          UGGAAAUCCAGAGUGGGUGAGGCCUGUCUGACCAUUUCUAGGCGACCCACUCUUGGUUUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012921
          miRBase ID: eca-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-551a
          Stem-loop Accession # MI0008768
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008231
          miRBase ID: ptr-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-551a
          Stem-loop Accession # MI0015045
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015997
          miRBase ID: ppy-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-551a
          Stem-loop Accession # MI0007827
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCAUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006419
          miRBase ID: mml-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03294054_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11396
          Ambion® Pre-miR™ miRNA Precursor : PM11396
          TaqMan™ MicroRNA Assay : 001519
          mirVana® miRNA inhibitor : MH11396
          mirVana® miRNA mimic : MC11396
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0