Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001519
          Assay Name: hsa-miR-551a
          miRBase Accession Number: MI0003556
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 3560695 - 3560790 [-] on Build GRCh38
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Species: Human, Dog, Gorilla gorilla, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001519
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000360, mir-551

          Mature miRNA Sequence:

          GCGACCCACUCUUGGUUUCCA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-551a MIMAT0003214
          Dog cfa-miR-551a MIMAT0009912
          Gorilla gorilla ggo-miR-551a MIMAT0024186
          Horse eca-miR-551a MIMAT0012921
          Pan troglodytes ptr-miR-551a MIMAT0008231
          Pongo pygmaeus ppy-miR-551a MIMAT0015997
          View More Rows
          Rhesus monkey mml-miR-551a MIMAT0006419

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-551a
          Stem-loop Accession # MI0003556
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCAGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAA
          Chromosome Location Chr. 1 - 3560695 - 3560790 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003214
          miRBase ID: hsa-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: Chr. 1 - 3560695 - 3560790 [-] on Build GRCh38

          Dog

          Stem-loop ID cfa-mir-551a
          Stem-loop Accession # MI0010411
          Stem-loop Sequence
          GGGGACUGCCAGGUGACCCUGGAAAUCCAGAGUGGGUAAGGCCUGUCUGAUUGUGUCUAGGCGACCCACUCUUGGUUUCCAUGGUUGCCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009912
          miRBase ID: cfa-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-551a
          Stem-loop Accession # MI0020733
          Stem-loop Sequence
          GGCUGCCAGAGGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAACCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024186
          miRBase ID: ggo-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-551a
          Stem-loop Accession # MI0012677
          Stem-loop Sequence
          UGGAAAUCCAGAGUGGGUGAGGCCUGUCUGACCAUUUCUAGGCGACCCACUCUUGGUUUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012921
          miRBase ID: eca-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-551a
          Stem-loop Accession # MI0008768
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008231
          miRBase ID: ptr-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-551a
          Stem-loop Accession # MI0015045
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCGUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015997
          miRBase ID: ppy-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-551a
          Stem-loop Accession # MI0007827
          Stem-loop Sequence
          GGGGACUGCCGGGUGACCCUGGAAAUCCAGAGUGGGUGGGGCCUGUCUGACCAUUUCUAGGCGACCCACUCUUGGUUUCCAGGGUUGCCCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006419
          miRBase ID: mml-miR-551a
          Mature miRNA Sequence: GCGACCCACUCUUGGUUUCCA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03294054_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11396
          Ambion® Pre-miR™ miRNA Precursor : PM11396
          mirVana® miRNA inhibitor : MH11396
          mirVana® miRNA mimic : MC11396
          TaqMan™ Advanced miRNA Assay : 478158_mir
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline