Search
Search

| Assay Name: | hsa-miR-877-5p |
|---|---|
| Stem-loop Accession Number: | MI0005561 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUAGAGGAGAUGGCGCAGGG |
| Species: | Human, Mouse, Rat, Bovine, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000392, mir-877 |
Mature miRNA Sequence: | GUAGAGGAGAUGGCGCAGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-877-5p | MIMAT0004949 |
| Mouse | mmu-miR-877-5p | MIMAT0004861 |
| Rat | rno-miR-877 | MIMAT0005285 |
| Bovine | bta-miR-877 | MIMAT0009381 |
| Pongo pygmaeus | ppy-miR-877 | MIMAT0016117 |
| Rhesus monkey | mml-miR-877-5p | MIMAT0006521 |