Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03308673_pri
Assay Name:
mmu-mir-877
Stem-loop ID:
mmu-mir-877
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.17: 35960730 - 35960814 [-] on Build GRCm38
Stem-loop Location:
Chr.17: 35960730 - 35960814 [-] on Build GRCm38
Mature miRNA ID:
mmu-miR-877-3p
mmu-miR-877-5p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03308673_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
58
Chromosome Location:
Chr. 17 - 35960536 - 35960536 [-] on 35960536 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-877-3p
MIMAT0004862
Mouse
mmu-miR-877-5p
MIMAT0004861
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-877
Stem-loop Accession #
MI0005553
Stem-loop Sequence
GUAGAGGAGAUGGCGCAGGGGACACAAGGUAGGCCUUGCGGGUCUGUGGACCCUUGGACAUGUGUCCUCUUCUCCCUCCUCCCAG
Chromosome Location
Chr. 17 - 35960730 - 35960814 [-] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0004862
miRBase ID: mmu-miR-877-3p
Mature miRNA Sequence: UGUCCUCUUCUCCCUCCUCCCA
Chromosome Location: Chr. 17 - 35960730 - 35960814 [-] on Build GRCm38
miRBase Accession #: MIMAT0004861
miRBase ID: mmu-miR-877-5p
Mature miRNA Sequence: GUAGAGGAGAUGGCGCAGGG
Chromosome Location: Chr. 17 - 35960730 - 35960814 [-] on Build GRCm38
Gene Family ID
MIPF0000392,
mir-877
Distance to Amplicon
169 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM13087
Ambion
®
Pre-miR
™
miRNA Precursor :
PM13087
TaqMan
™
MicroRNA Assay :
002548
mir
Vana
®
miRNA inhibitor :
MH13087
mir
Vana
®
miRNA mimic :
MC13087
TaqMan
™
Advanced miRNA Assay :
mmu482316_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12678
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12678
mir
Vana
®
miRNA inhibitor :
MH12678
mir
Vana
®
miRNA mimic :
MC12678
TaqMan
™
Advanced miRNA Assay :
478206_mir
TaqMan
™
Advanced miRNA Assay :
rno481273_mir
TaqMan
™
Advanced miRNA Assay :
mmu481273_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline