Search
Search

| Assay Name: | hsa-miR-128-2-5p |
|---|---|
| Stem-loop Accession Number: | MI0000727 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGCCGAUACACUGUACGAGA |
| Species: | Human, Dasypus novemcinctus, Guinea pig, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | GGGGGCCGAUACACUGUACGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-128-2-5p | MIMAT0031095 |
| Dasypus novemcinctus | dno-miR-128-2-5p | MIMAT0047623 |
| Guinea pig | cpo-miR-128-2-5p | MIMAT0046976 |
| Rabbit | ocu-miR-128b-5p | MIMAT0048212 |
| Rhesus monkey | mml-miR-128b-5p | MIMAT0026814 |