Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC27264
          Assay Name: hsa-miR-128-2-5p
          miRBase Accession Number: MI0000727
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 35744476 - 35744559 [+] on Build GRCh38
          Mature miRNA Sequence: GGGGGCCGAUACACUGUACGAGA
          Species: Human, Dasypus novemcinctus, Guinea pig, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC27264

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000048, mir-128

          Mature miRNA Sequence:

          GGGGGCCGAUACACUGUACGAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-128-2-5p MIMAT0031095
          Dasypus novemcinctus dno-miR-128-2-5p MIMAT0047623
          Guinea pig cpo-miR-128-2-5p MIMAT0046976
          Rabbit ocu-miR-128b-5p MIMAT0048212
          Rhesus monkey mml-miR-128b-5p MIMAT0026814

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-128-2
          Stem-loop Accession # MI0000727
          Stem-loop Sequence
          UGUGCAGUGGGAAGGGGGGCCGAUACACUGUACGAGAGUGAGUAGCAGGUCUCACAGUGAACCGGUCUCUUUCCCUACUGUGUC
          Chromosome Location Chr. 3 - 35744476 - 35744559 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0031095
          miRBase ID: hsa-miR-128-2-5p
          Mature miRNA Sequence: GGGGGCCGAUACACUGUACGAGA
          Chromosome Location: Chr. 3 - 35744476 - 35744559 [+] on Build GRCh38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-128-2
          Stem-loop Accession # MI0038971
          Stem-loop Sequence
          GGGGGCCGAUACACUGUACGAGAGUGAGUAGCAGGUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047623
          miRBase ID: dno-miR-128-2-5p
          Mature miRNA Sequence: GGGGGCCGAUACACUGUACGAGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-128-2
          Stem-loop Accession # MI0038630
          Stem-loop Sequence
          GGGGGCCGAUACACUGUACGAGAGUGAGUAGCAGGUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046976
          miRBase ID: cpo-miR-128-2-5p
          Mature miRNA Sequence: GGGGGCCGAUACACUGUACGAGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-128b
          Stem-loop Accession # MI0039282
          Stem-loop Sequence
          GGGGGCCGAUACACUGUACGAGAGUGAGUAGCAGGUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048212
          miRBase ID: ocu-miR-128b-5p
          Mature miRNA Sequence: GGGGGCCGAUACACUGUACGAGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-128b
          Stem-loop Accession # MI0007617
          Stem-loop Sequence
          UGUGCAGUGGGAAGGGGGGCCGAUACACUGUACGAGAGUGAGUAGCAGGUCUCACAGUGAACCGGUCUCUUUCCCUACUGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026814
          miRBase ID: mml-miR-128b-5p
          Mature miRNA Sequence: GGGGGCCGAUACACUGUACGAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303360_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM27264
          Ambion® Pre-miR™ miRNA Precursor : PM27264
          TaqMan™ MicroRNA Assay : 467291_mat
          mirVana® miRNA inhibitor : MH27264
          TaqMan™ Advanced miRNA Assay : 478684_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline